Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63917799-63918058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GH1
Entrez Gene ID
2688
Gene Name
growth hormone 1
Gene Aliases
GH, GH-N, GHB5, GHN, IGHD1B, hGH-N
UniGene
Hs.655229
Gene Chromosome Location
Chr.17: 63917193-63918852 on Build GRCh38
Transcripts
NM_000515.4 NM_022559.3 NM_022560.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3958649 c.282A>C p.Q94H Chr.17: 63918026-63918026 on Build GRCh38
3890168 c.369C>T p.F123F Chr.17: 63917847-63917847 on Build GRCh38
1610624 c.335G>A p.W112* Chr.17: 63917881-63917881 on Build GRCh38
4068663 c.269G>T p.R90M Chr.17: 63918039-63918039 on Build GRCh38
1522039 c.254C>T p.P85L Chr.17: 63918054-63918054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143833877 Chr.17: 63917826-63917826 on Build GRCh38
rs769287960 Chr.17: 63917931-63917931 on Build GRCh38
rs201518977 Chr.17: 63918017-63918017 on Build GRCh38
rs1045467832 Chr.17: 63918022-63918022 on Build GRCh38
rs748754824 Chr.17: 63917979-63917979 on Build GRCh38
rs797044450 Chr.17: 63918011-63918011 on Build GRCh38
rs763501141 Chr.17: 63917820-63917820 on Build GRCh38
rs6174 Chr.17: 63917902-63917902 on Build GRCh38
rs863223307 Chr.17: 63918012-63918012 on Build GRCh38
rs767954118 Chr.17: 63917947-63917947 on Build GRCh38
rs372776291 Chr.17: 63917864-63917864 on Build GRCh38
rs894190751 Chr.17: 63917892-63917892 on Build GRCh38
rs772906184 Chr.17: 63917938-63917938 on Build GRCh38
rs752055083 Chr.17: 63917835-63917835 on Build GRCh38
rs574412048 Chr.17: 63917985-63917985 on Build GRCh38
rs781058241 Chr.17: 63917964-63917964 on Build GRCh38
rs770588602 Chr.17: 63917981-63917981 on Build GRCh38
rs572761421 Chr.17: 63918029-63918029 on Build GRCh38
rs757586741 Chr.17: 63917960-63917960 on Build GRCh38
rs753248163 Chr.17: 63917949-63917949 on Build GRCh38
rs368216649 Chr.17: 63918002-63918002 on Build GRCh38
rs780577159 Chr.17: 63917833-63917833 on Build GRCh38
rs866454750 Chr.17: 63917852-63917852 on Build GRCh38
rs755141415 Chr.17: 63917837-63917837 on Build GRCh38
rs201524858 Chr.17: 63917828-63917828 on Build GRCh38
rs749511152 Chr.17: 63917900-63917900 on Build GRCh38
rs137853220 Chr.17: 63917909-63917909 on Build GRCh38
rs757500095 Chr.17: 63917908-63917908 on Build GRCh38
rs377027593 Chr.17: 63917870-63917870 on Build GRCh38
rs935822693 Chr.17: 63917994-63917994 on Build GRCh38
rs557973956 Chr.17: 63917913-63917913 on Build GRCh38
rs746829974 Chr.17: 63917963-63917963 on Build GRCh38
rs41295039 Chr.17: 63917940-63917940 on Build GRCh38
rs753904435 Chr.17: 63917959-63917959 on Build GRCh38
rs747651789 Chr.17: 63917929-63917929 on Build GRCh38
rs748100671 Chr.17: 63917966-63917966 on Build GRCh38
rs144610881 Chr.17: 63917871-63917871 on Build GRCh38
rs776608552 Chr.17: 63917925-63917925 on Build GRCh38
rs768500322 Chr.17: 63917922-63917922 on Build GRCh38
rs202020201 Chr.17: 63917991-63917991 on Build GRCh38
rs772711019 Chr.17: 63917973-63917973 on Build GRCh38
rs150761983 Chr.17: 63917888-63917888 on Build GRCh38
rs368658744 Chr.17: 63917857-63917857 on Build GRCh38
rs751860506 Chr.17: 63917877-63917877 on Build GRCh38
rs374067833 Chr.17: 63918007-63918007 on Build GRCh38
rs752738695 Chr.17: 63917885-63917885 on Build GRCh38
rs548995389 Chr.17: 63917859-63917859 on Build GRCh38
rs986174675 Chr.17: 63917975-63917975 on Build GRCh38
rs370785603 Chr.17: 63917829-63917829 on Build GRCh38
rs759328993 Chr.17: 63917995-63917996 on Build GRCh38
rs774071527 Chr.17: 63917960-63917960 on Build GRCh38
rs759888728 Chr.17: 63917988-63917988 on Build GRCh38
rs781692092 Chr.17: 63917884-63917884 on Build GRCh38
rs374293003 Chr.17: 63917992-63917992 on Build GRCh38
rs142396035 Chr.17: 63917866-63917866 on Build GRCh38
rs863223306 Chr.17: 63917989-63917989 on Build GRCh38
rs373958758 Chr.17: 63917846-63917846 on Build GRCh38
rs748188371 Chr.17: 63917847-63917847 on Build GRCh38
rs775630395 Chr.17: 63917861-63917861 on Build GRCh38
rs137882374 Chr.17: 63917853-63917853 on Build GRCh38
rs41295037 Chr.17: 63917945-63917945 on Build GRCh38
rs541390525 Chr.17: 63917940-63917940 on Build GRCh38
rs200587202 Chr.17: 63917941-63917941 on Build GRCh38
rs71640278 Chr.17: 63917883-63917883 on Build GRCh38
rs756208586 Chr.17: 63917951-63917951 on Build GRCh38
rs746780250 Chr.17: 63917921-63917921 on Build GRCh38
rs749382898 Chr.17: 63917984-63917984 on Build GRCh38
rs915740374 Chr.17: 63918006-63918006 on Build GRCh38
rs200503313 Chr.17: 63917939-63917939 on Build GRCh38
rs886053236 Chr.17: 63917907-63917907 on Build GRCh38
rs71640277 Chr.17: 63918016-63918016 on Build GRCh38
rs769659274 Chr.17: 63917972-63917972 on Build GRCh38
rs774605655 Chr.17: 63917943-63917943 on Build GRCh38
rs139163836 Chr.17: 63917889-63917889 on Build GRCh38
rs765595617 Chr.17: 63918003-63918003 on Build GRCh38
rs367960425 Chr.17: 63918009-63918009 on Build GRCh38
rs753333968 Chr.17: 63917825-63917825 on Build GRCh38
rs863223310 Chr.17: 63918015-63918015 on Build GRCh38
rs766715590 Chr.17: 63917875-63917875 on Build GRCh38
rs753995450 Chr.17: 63917997-63917997 on Build GRCh38
rs776036784 Chr.17: 63917867-63917867 on Build GRCh38
rs372156665 Chr.17: 63918004-63918004 on Build GRCh38

View Full Product Details