Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46723379-46723606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2
Entrez Gene ID
2147
Gene Name
coagulation factor II, thrombin
Gene Aliases
PT, RPRGL2, THPH1
UniGene
Hs.655207
Gene Chromosome Location
Chr.11: 46719166-46739508 on Build GRCh38
Transcripts
NM_000506.4 NM_001311257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1980874 c.495G>A p.T165T Chr.11: 46723454-46723454 on Build GRCh38
1980873 c.459G>A p.Q153Q Chr.11: 46723418-46723418 on Build GRCh38
1980876 c.497G>T p.G166V Chr.11: 46723456-46723456 on Build GRCh38
1980875 c.496G>T p.G166* Chr.11: 46723455-46723455 on Build GRCh38
7291955 c.481G>A p.D161N Chr.11: 46723440-46723440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778851027 Chr.11: 46723521-46723521 on Build GRCh38
rs775320386 Chr.11: 46723519-46723520 on Build GRCh38
rs371627939 Chr.11: 46723483-46723483 on Build GRCh38
rs113273575 Chr.11: 46723520-46723520 on Build GRCh38
rs375002682 Chr.11: 46723562-46723562 on Build GRCh38
rs761400213 Chr.11: 46723522-46723523 on Build GRCh38
rs774792190 Chr.11: 46723476-46723476 on Build GRCh38
rs773232576 Chr.11: 46723458-46723458 on Build GRCh38
rs777855413 Chr.11: 46723540-46723540 on Build GRCh38
rs900863560 Chr.11: 46723438-46723438 on Build GRCh38
rs752361229 Chr.11: 46723485-46723485 on Build GRCh38
rs369468882 Chr.11: 46723460-46723460 on Build GRCh38
rs371923575 Chr.11: 46723409-46723409 on Build GRCh38
rs568347016 Chr.11: 46723436-46723436 on Build GRCh38
rs977169393 Chr.11: 46723541-46723541 on Build GRCh38
rs764154260 Chr.11: 46723421-46723421 on Build GRCh38
rs768825889 Chr.11: 46723431-46723431 on Build GRCh38
rs3136452 Chr.11: 46723439-46723439 on Build GRCh38
rs149943083 Chr.11: 46723410-46723410 on Build GRCh38
rs201846578 Chr.11: 46723545-46723545 on Build GRCh38
rs775023243 Chr.11: 46723558-46723558 on Build GRCh38
rs760307675 Chr.11: 46723493-46723493 on Build GRCh38
rs757232176 Chr.11: 46723511-46723511 on Build GRCh38
rs773827956 Chr.11: 46723521-46723522 on Build GRCh38
rs781116672 Chr.11: 46723518-46723519 on Build GRCh38
rs758192520 Chr.11: 46723530-46723530 on Build GRCh38
rs753359568 Chr.11: 46723509-46723509 on Build GRCh38
rs772848999 Chr.11: 46723523-46723524 on Build GRCh38
rs144857547 Chr.11: 46723454-46723454 on Build GRCh38
rs749059497 Chr.11: 46723520-46723521 on Build GRCh38
rs767818793 Chr.11: 46723484-46723484 on Build GRCh38
rs387906522 Chr.11: 46723421-46723422 on Build GRCh38
rs994858528 Chr.11: 46723514-46723514 on Build GRCh38
rs780567837 Chr.11: 46723469-46723469 on Build GRCh38
rs769969150 Chr.11: 46723539-46723539 on Build GRCh38
rs779756296 Chr.11: 46723535-46723535 on Build GRCh38
rs3136454 Chr.11: 46723538-46723538 on Build GRCh38
rs5896 Chr.11: 46723453-46723453 on Build GRCh38
rs1026722245 Chr.11: 46723452-46723452 on Build GRCh38
rs771528049 Chr.11: 46723557-46723557 on Build GRCh38
rs376151472 Chr.11: 46723523-46723523 on Build GRCh38
rs778621867 Chr.11: 46723419-46723419 on Build GRCh38
rs374766750 Chr.11: 46723440-46723440 on Build GRCh38
rs763643404 Chr.11: 46723498-46723498 on Build GRCh38

View Full Product Details