Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77949086-77949359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUBP1
Entrez Gene ID
8880
Gene Name
far upstream element binding protein 1
Gene Aliases
FBP, FUBP, hDH V
UniGene
Hs.567380
Gene Chromosome Location
Chr.1: 77944055-77979494 on Build GRCh38
Transcripts
NM_001303433.1 NM_003902.4 NR_130152.1 XM_011542391.2 XM_011542392.2 XM_011542393.2 XM_017002739.1 XM_017002740.1 XM_017002741.1 XM_017002742.1 XM_017002743.1 XM_017002744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6957210 c.1926+1G>A p.? Chr.1: 77949154-77949154 on Build GRCh38
4009984 c.1907A>C p.Q636P Chr.1: 77949174-77949174 on Build GRCh38
6931589 c.1805G>A p.G602E Chr.1: 77949276-77949276 on Build GRCh38
1716767 c.1855_1856delTA p.Y619fs*1 Chr.1: 77949225-77949226 on Build GRCh38
6940278 c.1876C>T p.P626S Chr.1: 77949268-77949268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779496471 Chr.1: 77949327-77949327 on Build GRCh38
rs757124726 Chr.1: 77949314-77949314 on Build GRCh38
rs569201989 Chr.1: 77949311-77949311 on Build GRCh38
rs751256080 Chr.1: 77949313-77949313 on Build GRCh38
rs752653413 Chr.1: 77949275-77949275 on Build GRCh38
rs1011325852 Chr.1: 77949315-77949315 on Build GRCh38
rs997164355 Chr.1: 77949178-77949178 on Build GRCh38
rs776768914 Chr.1: 77949304-77949305 on Build GRCh38
rs200862215 Chr.1: 77949123-77949123 on Build GRCh38
rs776042359 Chr.1: 77949162-77949162 on Build GRCh38
rs571394258 Chr.1: 77949126-77949126 on Build GRCh38
rs200421915 Chr.1: 77949197-77949197 on Build GRCh38
rs200035916 Chr.1: 77949273-77949273 on Build GRCh38
rs762232309 Chr.1: 77949305-77949306 on Build GRCh38
rs150838067 Chr.1: 77949195-77949195 on Build GRCh38
rs374511552 Chr.1: 77949326-77949326 on Build GRCh38
rs368516501 Chr.1: 77949329-77949329 on Build GRCh38
rs567186426 Chr.1: 77949247-77949247 on Build GRCh38
rs769055797 Chr.1: 77949189-77949189 on Build GRCh38
rs539167724 Chr.1: 77949257-77949257 on Build GRCh38
rs368096513 Chr.1: 77949218-77949218 on Build GRCh38
rs902092198 Chr.1: 77949164-77949164 on Build GRCh38
rs763409195 Chr.1: 77949169-77949169 on Build GRCh38
rs901160335 Chr.1: 77949279-77949279 on Build GRCh38
rs762248194 Chr.1: 77949207-77949207 on Build GRCh38
rs984183819 Chr.1: 77949139-77949139 on Build GRCh38
rs758194583 Chr.1: 77949118-77949118 on Build GRCh38
rs777472929 Chr.1: 77949122-77949122 on Build GRCh38
rs587778376 Chr.1: 77949280-77949280 on Build GRCh38
rs370174403 Chr.1: 77949281-77949281 on Build GRCh38
rs745329616 Chr.1: 77949141-77949141 on Build GRCh38
rs769449219 Chr.1: 77949158-77949158 on Build GRCh38
rs780941453 Chr.1: 77949318-77949318 on Build GRCh38
rs367613192 Chr.1: 77949150-77949150 on Build GRCh38

View Full Product Details