Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6214350-6214607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
341668 c.2522A>T p.Y841F Chr.11: 6214488-6214488 on Build GRCh38
2038953 c.2465A>T p.E822V Chr.11: 6214545-6214545 on Build GRCh38
6277230 c.2588C>A p.S863Y Chr.11: 6214422-6214422 on Build GRCh38
5495690 c.2538C>T p.G846G Chr.11: 6214472-6214472 on Build GRCh38
6361861 c.2590G>T p.D864Y Chr.11: 6214420-6214420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765897198 Chr.11: 6214383-6214383 on Build GRCh38
rs752434654 Chr.11: 6214482-6214482 on Build GRCh38
rs773866778 Chr.11: 6214515-6214515 on Build GRCh38
rs759141820 Chr.11: 6214477-6214477 on Build GRCh38
rs745873221 Chr.11: 6214417-6214417 on Build GRCh38
rs755678121 Chr.11: 6214483-6214483 on Build GRCh38
rs375013443 Chr.11: 6214454-6214454 on Build GRCh38
rs267603071 Chr.11: 6214502-6214502 on Build GRCh38
rs779696631 Chr.11: 6214583-6214583 on Build GRCh38
rs779054701 Chr.11: 6214511-6214511 on Build GRCh38
rs367763962 Chr.11: 6214514-6214514 on Build GRCh38
rs149153168 Chr.11: 6214425-6214425 on Build GRCh38
rs147815418 Chr.11: 6214510-6214510 on Build GRCh38
rs112595124 Chr.11: 6214526-6214526 on Build GRCh38
rs768704295 Chr.11: 6214530-6214530 on Build GRCh38
rs576094611 Chr.11: 6214527-6214527 on Build GRCh38
rs766892896 Chr.11: 6214552-6214553 on Build GRCh38
rs776932941 Chr.11: 6214464-6214464 on Build GRCh38
rs762234135 Chr.11: 6214550-6214550 on Build GRCh38
rs765667584 Chr.11: 6214469-6214469 on Build GRCh38
rs975885209 Chr.11: 6214565-6214565 on Build GRCh38
rs764347392 Chr.11: 6214378-6214378 on Build GRCh38
rs117940741 Chr.11: 6214426-6214426 on Build GRCh38
rs760780932 Chr.11: 6214428-6214428 on Build GRCh38
rs74735527 Chr.11: 6214380-6214380 on Build GRCh38
rs761543473 Chr.11: 6214546-6214547 on Build GRCh38
rs745773818 Chr.11: 6214513-6214513 on Build GRCh38
rs777729283 Chr.11: 6214489-6214489 on Build GRCh38
rs925661853 Chr.11: 6214396-6214396 on Build GRCh38
rs374303574 Chr.11: 6214385-6214385 on Build GRCh38
rs371845795 Chr.11: 6214429-6214429 on Build GRCh38
rs755984775 Chr.11: 6214395-6214395 on Build GRCh38
rs116653213 Chr.11: 6214388-6214388 on Build GRCh38
rs766913546 Chr.11: 6214480-6214480 on Build GRCh38
rs1001793443 Chr.11: 6214431-6214431 on Build GRCh38
rs773757036 Chr.11: 6214571-6214571 on Build GRCh38
rs200682955 Chr.11: 6214412-6214412 on Build GRCh38
rs143007514 Chr.11: 6214387-6214387 on Build GRCh38
rs755782401 Chr.11: 6214554-6214554 on Build GRCh38
rs779144678 Chr.11: 6214415-6214415 on Build GRCh38
rs762322159 Chr.11: 6214468-6214468 on Build GRCh38
rs776986122 Chr.11: 6214538-6214538 on Build GRCh38
rs749265378 Chr.11: 6214404-6214404 on Build GRCh38
rs751637423 Chr.11: 6214379-6214379 on Build GRCh38
rs374453791 Chr.11: 6214401-6214401 on Build GRCh38
rs747277340 Chr.11: 6214529-6214529 on Build GRCh38
rs999441433 Chr.11: 6214381-6214381 on Build GRCh38
rs750927334 Chr.11: 6214476-6214476 on Build GRCh38
rs377292028 Chr.11: 6214577-6214577 on Build GRCh38

View Full Product Details