Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14408834-14409095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX39A
Entrez Gene ID
10212
Gene Name
DEAD-box helicase 39A
Gene Aliases
BAT1, BAT1L, DDX39, DDXL, URH49
UniGene
Hs.311609
Gene Chromosome Location
Chr.19: 14408798-14419383 on Build GRCh38
Transcripts
NM_005804.3 NR_046366.2 XM_006722606.3 XM_011527620.1 XM_011527621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3712730 c.1269C>T p.I423I Chr.19: 14408951-14408951 on Build GRCh38
3822108 c.1267A>C p.I423L Chr.19: 14409037-14409037 on Build GRCh38
4131279 c.1280G>T p.R427L Chr.19: 14408940-14408940 on Build GRCh38
5936448 c.1270G>A p.E424K Chr.19: 14408950-14408950 on Build GRCh38
4074795 c.1227T>C p.N409N Chr.19: 14409077-14409077 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199544696 Chr.19: 14408929-14408929 on Build GRCh38
rs760726811 Chr.19: 14408990-14408990 on Build GRCh38
rs752148704 Chr.19: 14408955-14408955 on Build GRCh38
rs75381848 Chr.19: 14408871-14408871 on Build GRCh38
rs753145012 Chr.19: 14408942-14408942 on Build GRCh38
rs761600377 Chr.19: 14408910-14408910 on Build GRCh38
rs375322648 Chr.19: 14408900-14408900 on Build GRCh38
rs879047758 Chr.19: 14408953-14408953 on Build GRCh38
rs199718078 Chr.19: 14409032-14409032 on Build GRCh38
rs753057074 Chr.19: 14408885-14408885 on Build GRCh38
rs779507230 Chr.19: 14408904-14408904 on Build GRCh38
rs777978127 Chr.19: 14408951-14408951 on Build GRCh38
rs904163542 Chr.19: 14408884-14408884 on Build GRCh38
rs150156465 Chr.19: 14408928-14408928 on Build GRCh38
rs377622562 Chr.19: 14408988-14408988 on Build GRCh38
rs757456778 Chr.19: 14408895-14408895 on Build GRCh38
rs148589157 Chr.19: 14409053-14409053 on Build GRCh38
rs112255397 Chr.19: 14409036-14409036 on Build GRCh38
rs370520494 Chr.19: 14408899-14408899 on Build GRCh38
rs768282153 Chr.19: 14408907-14408907 on Build GRCh38
rs758743399 Chr.19: 14408946-14408946 on Build GRCh38
rs200664433 Chr.19: 14408939-14408939 on Build GRCh38
rs745394150 Chr.19: 14408902-14408902 on Build GRCh38
rs374486556 Chr.19: 14408917-14408917 on Build GRCh38
rs371430739 Chr.19: 14408909-14408909 on Build GRCh38
rs1020240189 Chr.19: 14408959-14408959 on Build GRCh38
rs368642237 Chr.19: 14408918-14408918 on Build GRCh38
rs539797015 Chr.19: 14409026-14409026 on Build GRCh38
rs759650855 Chr.19: 14409043-14409043 on Build GRCh38
rs370976136 Chr.19: 14409005-14409005 on Build GRCh38
rs749006416 Chr.19: 14408906-14408906 on Build GRCh38
rs763054878 Chr.19: 14409068-14409068 on Build GRCh38
rs550997246 Chr.19: 14408966-14408966 on Build GRCh38
rs778493441 Chr.19: 14408971-14408971 on Build GRCh38
rs867431700 Chr.19: 14408961-14408961 on Build GRCh38
rs760503836 Chr.19: 14408914-14408914 on Build GRCh38
rs201988573 Chr.19: 14408991-14408991 on Build GRCh38
rs373517700 Chr.19: 14409037-14409037 on Build GRCh38
rs188739994 Chr.19: 14408903-14408903 on Build GRCh38
rs754959688 Chr.19: 14408935-14408936 on Build GRCh38
rs748294745 Chr.19: 14408987-14408987 on Build GRCh38
rs771931831 Chr.19: 14408911-14408911 on Build GRCh38
rs768300363 Chr.19: 14408970-14408970 on Build GRCh38
rs778651208 Chr.19: 14408954-14408955 on Build GRCh38
rs984520529 Chr.19: 14409024-14409024 on Build GRCh38
rs766154814 Chr.19: 14408900-14408913 on Build GRCh38
rs748736583 Chr.19: 14408968-14408968 on Build GRCh38
rs559768023 Chr.19: 14408872-14408872 on Build GRCh38
rs764587694 Chr.19: 14409073-14409073 on Build GRCh38
rs369602896 Chr.19: 14408958-14408958 on Build GRCh38
rs753774578 Chr.19: 14408910-14408923 on Build GRCh38
rs574444721 Chr.19: 14409045-14409045 on Build GRCh38
rs371485435 Chr.19: 14408986-14408986 on Build GRCh38
rs374521557 Chr.19: 14408987-14408987 on Build GRCh38
rs773149157 Chr.19: 14408913-14408913 on Build GRCh38

View Full Product Details