Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87375485-87375710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
UniGene
Hs.125039
Gene Chromosome Location
Chr.7: 87345635-87399796 on Build GRCh38
Transcripts
NM_001143935.1 NM_021151.3 XM_011516337.2 XM_017012370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7183052 c.677A>G p.K226R Chr.7: 87375652-87375652 on Build GRCh38
3032493 c.734G>A p.R245Q Chr.7: 87375709-87375709 on Build GRCh38
1092557 c.678G>T p.K226N Chr.7: 87375653-87375653 on Build GRCh38
5510229 c.666_667delAT p.I224fs*6 Chr.7: 87375641-87375642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777097001 Chr.7: 87375599-87375600 on Build GRCh38
rs756910824 Chr.7: 87375629-87375629 on Build GRCh38
rs955787651 Chr.7: 87375554-87375554 on Build GRCh38
rs188365193 Chr.7: 87375594-87375594 on Build GRCh38
rs1031837115 Chr.7: 87375537-87375537 on Build GRCh38
rs202220049 Chr.7: 87375685-87375685 on Build GRCh38
rs374185276 Chr.7: 87375649-87375649 on Build GRCh38
rs760584984 Chr.7: 87375595-87375595 on Build GRCh38
rs557853080 Chr.7: 87375638-87375638 on Build GRCh38
rs996243707 Chr.7: 87375651-87375651 on Build GRCh38
rs1013921239 Chr.7: 87375590-87375590 on Build GRCh38
rs199966683 Chr.7: 87375648-87375648 on Build GRCh38
rs1028693396 Chr.7: 87375652-87375652 on Build GRCh38
rs773304654 Chr.7: 87375688-87375688 on Build GRCh38
rs146451977 Chr.7: 87375661-87375661 on Build GRCh38
rs748387087 Chr.7: 87375678-87375678 on Build GRCh38
rs891989116 Chr.7: 87375556-87375556 on Build GRCh38
rs769040801 Chr.7: 87375667-87375667 on Build GRCh38
rs753563567 Chr.7: 87375607-87375607 on Build GRCh38
rs375390435 Chr.7: 87375524-87375526 on Build GRCh38
rs763937408 Chr.7: 87375596-87375596 on Build GRCh38
rs565632583 Chr.7: 87375579-87375579 on Build GRCh38
rs781490447 Chr.7: 87375675-87375675 on Build GRCh38

View Full Product Details