Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43365179-43365442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL1
Entrez Gene ID
64834
Gene Name
ELOVL fatty acid elongase 1
Gene Aliases
CGI-88, Ssc1
UniGene
Hs.25597
Gene Chromosome Location
Chr.1: 43363397-43368074 on Build GRCh38
Transcripts
NM_001256399.1 NM_001256401.1 NM_001256402.1 NM_022821.3 NR_046117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909763 c.52C>T p.R18W Chr.1: 43365371-43365371 on Build GRCh38
5911311 c.134T>C p.L45P Chr.1: 43365289-43365289 on Build GRCh38
1205218 c.143G>A p.R48H Chr.1: 43365280-43365280 on Build GRCh38
4534354 c.207G>A p.L69L Chr.1: 43365216-43365216 on Build GRCh38
3489894 c.175C>T p.R59C Chr.1: 43365248-43365248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769555812 Chr.1: 43365247-43365247 on Build GRCh38
rs199630809 Chr.1: 43365210-43365210 on Build GRCh38
rs746362044 Chr.1: 43365207-43365207 on Build GRCh38
rs142106087 Chr.1: 43365295-43365295 on Build GRCh38
rs201270403 Chr.1: 43365266-43365266 on Build GRCh38
rs376128835 Chr.1: 43365419-43365419 on Build GRCh38
rs766301849 Chr.1: 43365276-43365276 on Build GRCh38
rs754465886 Chr.1: 43365280-43365280 on Build GRCh38
rs775452198 Chr.1: 43365220-43365220 on Build GRCh38
rs750748004 Chr.1: 43365323-43365323 on Build GRCh38
rs747133232 Chr.1: 43365339-43365339 on Build GRCh38
rs772913569 Chr.1: 43365374-43365374 on Build GRCh38
rs763673976 Chr.1: 43365417-43365417 on Build GRCh38
rs143854596 Chr.1: 43365244-43365244 on Build GRCh38
rs755693311 Chr.1: 43365300-43365300 on Build GRCh38
rs945916093 Chr.1: 43365405-43365406 on Build GRCh38
rs773003850 Chr.1: 43365265-43365265 on Build GRCh38
rs772569715 Chr.1: 43365209-43365209 on Build GRCh38
rs751073844 Chr.1: 43365278-43365278 on Build GRCh38
rs774329407 Chr.1: 43365389-43365389 on Build GRCh38
rs770819986 Chr.1: 43365383-43365383 on Build GRCh38
rs767218638 Chr.1: 43365281-43365281 on Build GRCh38
rs913667405 Chr.1: 43365214-43365214 on Build GRCh38
rs762775512 Chr.1: 43365382-43365382 on Build GRCh38
rs769038287 Chr.1: 43365221-43365221 on Build GRCh38
rs776970798 Chr.1: 43365235-43365235 on Build GRCh38
rs752347198 Chr.1: 43365283-43365283 on Build GRCh38
rs761758758 Chr.1: 43365245-43365245 on Build GRCh38
rs765129030 Chr.1: 43365257-43365257 on Build GRCh38
rs752149010 Chr.1: 43365410-43365410 on Build GRCh38
rs937682878 Chr.1: 43365320-43365320 on Build GRCh38
rs868431896 Chr.1: 43365404-43365404 on Build GRCh38
rs200393447 Chr.1: 43365357-43365357 on Build GRCh38
rs75531338 Chr.1: 43365318-43365318 on Build GRCh38
rs758858435 Chr.1: 43365327-43365327 on Build GRCh38
rs374025661 Chr.1: 43365396-43365396 on Build GRCh38
rs780578381 Chr.1: 43365328-43365328 on Build GRCh38
rs1054323045 Chr.1: 43365388-43365388 on Build GRCh38
rs767024087 Chr.1: 43365398-43365398 on Build GRCh38
rs370510277 Chr.1: 43365341-43365341 on Build GRCh38
rs370638221 Chr.1: 43365347-43365347 on Build GRCh38
rs781354323 Chr.1: 43365345-43365345 on Build GRCh38

View Full Product Details