Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89185257-89185510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR1
Entrez Gene ID
2569
Gene Name
gamma-aminobutyric acid type A receptor rho1 subunit
Gene Aliases
-
UniGene
Hs.99927
Gene Chromosome Location
Chr.6: 89177504-89231288 on Build GRCh38
Transcripts
NM_001256703.1 NM_001256704.1 NM_001267582.1 NM_002042.4 XM_017010689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082449 c.726G>A p.R242R Chr.6: 89185380-89185380 on Build GRCh38
7127953 c.656-1G>C p.? Chr.6: 89185451-89185451 on Build GRCh38
5890174 c.690G>A p.W230* Chr.6: 89185416-89185416 on Build GRCh38
4758660 c.752A>G p.Q251R Chr.6: 89185354-89185354 on Build GRCh38
1622019 c.726G>T p.R242R Chr.6: 89185380-89185380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942995139 Chr.6: 89185477-89185477 on Build GRCh38
rs753178099 Chr.6: 89185330-89185330 on Build GRCh38
rs759670607 Chr.6: 89185427-89185427 on Build GRCh38
rs364936 Chr.6: 89185488-89185488 on Build GRCh38
rs748672519 Chr.6: 89185378-89185378 on Build GRCh38
rs755524351 Chr.6: 89185343-89185343 on Build GRCh38
rs561725817 Chr.6: 89185467-89185467 on Build GRCh38
rs750271212 Chr.6: 89185479-89185479 on Build GRCh38
rs765071223 Chr.6: 89185282-89185283 on Build GRCh38
rs370866905 Chr.6: 89185381-89185381 on Build GRCh38
rs200602822 Chr.6: 89185382-89185382 on Build GRCh38
rs751266463 Chr.6: 89185487-89185487 on Build GRCh38
rs758117017 Chr.6: 89185326-89185326 on Build GRCh38
rs550894841 Chr.6: 89185486-89185486 on Build GRCh38
rs377734727 Chr.6: 89185469-89185469 on Build GRCh38
rs761458075 Chr.6: 89185294-89185294 on Build GRCh38
rs406462 Chr.6: 89185483-89185483 on Build GRCh38
rs774412746 Chr.6: 89185408-89185408 on Build GRCh38
rs781739684 Chr.6: 89185360-89185360 on Build GRCh38
rs752879906 Chr.6: 89185463-89185464 on Build GRCh38
rs554779889 Chr.6: 89185466-89185467 on Build GRCh38
rs377153156 Chr.6: 89185317-89185317 on Build GRCh38
rs751178900 Chr.6: 89185338-89185338 on Build GRCh38
rs749994267 Chr.6: 89185320-89185320 on Build GRCh38
rs866428923 Chr.6: 89185335-89185335 on Build GRCh38
rs765722653 Chr.6: 89185442-89185442 on Build GRCh38
rs201745195 Chr.6: 89185405-89185405 on Build GRCh38
rs1051311188 Chr.6: 89185386-89185386 on Build GRCh38
rs772154225 Chr.6: 89185430-89185430 on Build GRCh38
rs1055557826 Chr.6: 89185387-89185387 on Build GRCh38
rs762234427 Chr.6: 89185333-89185333 on Build GRCh38
rs554380353 Chr.6: 89185354-89185354 on Build GRCh38
rs764917011 Chr.6: 89185478-89185478 on Build GRCh38

View Full Product Details