Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48375226-48375487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN2
Entrez Gene ID
3344
Gene Name
forkhead box N2
Gene Aliases
HTLF
UniGene
Hs.468478
Gene Chromosome Location
Chr.2: 48314640-48379295 on Build GRCh38
Transcripts
NM_002158.3 XM_005264282.2 XM_005264283.2 XM_006712001.3 XM_006712002.3 XM_011532808.2 XM_017003956.1 XM_017003957.1 XM_017003958.1 XM_017003959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207441 c.1157G>A p.R386Q Chr.2: 48375304-48375304 on Build GRCh38
7181211 c.1152G>A p.K384K Chr.2: 48375299-48375299 on Build GRCh38
5669110 c.1186G>T p.E396* Chr.2: 48375333-48375333 on Build GRCh38
721717 c.1293A>G p.K431K Chr.2: 48375440-48375440 on Build GRCh38
4583535 c.1115G>A p.C372Y Chr.2: 48375262-48375262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773885405 Chr.2: 48375415-48375415 on Build GRCh38
rs777291785 Chr.2: 48375394-48375394 on Build GRCh38
rs755914589 Chr.2: 48375455-48375455 on Build GRCh38
rs748357224 Chr.2: 48375362-48375362 on Build GRCh38
rs761311561 Chr.2: 48375277-48375277 on Build GRCh38
rs917524043 Chr.2: 48375428-48375428 on Build GRCh38
rs762558339 Chr.2: 48375288-48375288 on Build GRCh38
rs780937696 Chr.2: 48375391-48375391 on Build GRCh38
rs756108709 Chr.2: 48375313-48375313 on Build GRCh38
rs750200540 Chr.2: 48375304-48375304 on Build GRCh38
rs748349616 Chr.2: 48375262-48375262 on Build GRCh38
rs779009569 Chr.2: 48375347-48375347 on Build GRCh38
rs141186693 Chr.2: 48375379-48375379 on Build GRCh38
rs775209088 Chr.2: 48375283-48375283 on Build GRCh38
rs924899951 Chr.2: 48375435-48375435 on Build GRCh38
rs955015109 Chr.2: 48375353-48375353 on Build GRCh38
rs745810950 Chr.2: 48375433-48375433 on Build GRCh38
rs144245594 Chr.2: 48375318-48375318 on Build GRCh38
rs755030343 Chr.2: 48375326-48375326 on Build GRCh38
rs528024655 Chr.2: 48375420-48375420 on Build GRCh38
rs761423435 Chr.2: 48375417-48375417 on Build GRCh38
rs779967615 Chr.2: 48375316-48375316 on Build GRCh38
rs1007175493 Chr.2: 48375282-48375282 on Build GRCh38
rs561146723 Chr.2: 48375456-48375456 on Build GRCh38
rs760072258 Chr.2: 48375398-48375398 on Build GRCh38
rs768712035 Chr.2: 48375257-48375257 on Build GRCh38
rs545664545 Chr.2: 48375270-48375270 on Build GRCh38
rs1055769781 Chr.2: 48375383-48375383 on Build GRCh38
rs143561178 Chr.2: 48375432-48375432 on Build GRCh38
rs1019361333 Chr.2: 48375403-48375403 on Build GRCh38
rs774446939 Chr.2: 48375259-48375259 on Build GRCh38
rs751194460 Chr.2: 48375300-48375300 on Build GRCh38
rs774849644 Chr.2: 48375247-48375247 on Build GRCh38
rs772354846 Chr.2: 48375267-48375267 on Build GRCh38
rs201814453 Chr.2: 48375289-48375289 on Build GRCh38
rs767260719 Chr.2: 48375303-48375303 on Build GRCh38
rs996793687 Chr.2: 48375299-48375299 on Build GRCh38
rs778887681 Chr.2: 48375434-48375435 on Build GRCh38
rs577804315 Chr.2: 48375256-48375256 on Build GRCh38
rs189152161 Chr.2: 48375374-48375374 on Build GRCh38
rs756898015 Chr.2: 48375302-48375302 on Build GRCh38
rs761175861 Chr.2: 48375314-48375314 on Build GRCh38
rs377347494 Chr.2: 48375378-48375378 on Build GRCh38
rs763547896 Chr.2: 48375411-48375411 on Build GRCh38
rs772220578 Chr.2: 48375359-48375359 on Build GRCh38
rs77721637 Chr.2: 48375294-48375294 on Build GRCh38
rs750007998 Chr.2: 48375444-48375444 on Build GRCh38
rs953114067 Chr.2: 48375251-48375251 on Build GRCh38

View Full Product Details