Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75028619-75028866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUPD1
Entrez Gene ID
338599
Gene Name
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases
DUSP27, FMDSP
UniGene
Hs.535090
Gene Chromosome Location
Chr.10: 75026427-75073827 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Gene Symbol
KAT6B
Entrez Gene ID
23522
Gene Name
lysine acetyltransferase 6B
Gene Aliases
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
UniGene
Hs.35758
Gene Chromosome Location
Chr.10: 74824927-75032623 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1965153 c.3984A>G p.E1328E Chr.10: 75028808-75028808 on Build GRCh38
4492102 c.3910C>T p.P1304S Chr.10: 75028734-75028734 on Build GRCh38
7145527 c.3882G>C p.E1294D Chr.10: 75028706-75028706 on Build GRCh38
3807734 c.3961C>G p.Q1321E Chr.10: 75028785-75028785 on Build GRCh38
427942 c.3916A>T p.R1306W Chr.10: 75028740-75028740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778203913 Chr.10: 75028718-75028718 on Build GRCh38
rs141935235 Chr.10: 75028652-75028652 on Build GRCh38
rs778493952 Chr.10: 75028738-75028738 on Build GRCh38
rs756421689 Chr.10: 75028679-75028679 on Build GRCh38
rs766315089 Chr.10: 75028661-75028661 on Build GRCh38
rs573655531 Chr.10: 75028796-75028796 on Build GRCh38
rs150339818 Chr.10: 75028781-75028781 on Build GRCh38
rs759590847 Chr.10: 75028792-75028792 on Build GRCh38
rs776501920 Chr.10: 75028778-75028778 on Build GRCh38
rs151215354 Chr.10: 75028766-75028766 on Build GRCh38
rs886041987 Chr.10: 75028742-75028742 on Build GRCh38
rs376367047 Chr.10: 75028676-75028676 on Build GRCh38
rs368121049 Chr.10: 75028663-75028663 on Build GRCh38
rs577563268 Chr.10: 75028664-75028664 on Build GRCh38
rs371747862 Chr.10: 75028671-75028671 on Build GRCh38
rs201742830 Chr.10: 75028724-75028724 on Build GRCh38
rs997181148 Chr.10: 75028821-75028821 on Build GRCh38
rs978272228 Chr.10: 75028653-75028653 on Build GRCh38
rs199470475 Chr.10: 75028716-75028716 on Build GRCh38
rs753136232 Chr.10: 75028801-75028801 on Build GRCh38
rs539703139 Chr.10: 75028648-75028648 on Build GRCh38
rs764565963 Chr.10: 75028818-75028818 on Build GRCh38
rs756960480 Chr.10: 75028767-75028767 on Build GRCh38
rs770602078 Chr.10: 75028732-75028732 on Build GRCh38
rs754223525 Chr.10: 75028824-75028824 on Build GRCh38
rs201959220 Chr.10: 75028752-75028752 on Build GRCh38
rs142230616 Chr.10: 75028755-75028755 on Build GRCh38
rs770307489 Chr.10: 75028665-75028665 on Build GRCh38
rs757599670 Chr.10: 75028828-75028828 on Build GRCh38
rs138030591 Chr.10: 75028786-75028786 on Build GRCh38
rs777272076 Chr.10: 75028658-75028658 on Build GRCh38
rs863224883 Chr.10: 75028786-75028787 on Build GRCh38
rs766620893 Chr.10: 75028782-75028782 on Build GRCh38
rs572647460 Chr.10: 75028670-75028670 on Build GRCh38
rs140301566 Chr.10: 75028776-75028776 on Build GRCh38
rs199470474 Chr.10: 75028701-75028701 on Build GRCh38
rs201891685 Chr.10: 75028816-75028816 on Build GRCh38
rs778602696 Chr.10: 75028835-75028835 on Build GRCh38
rs768503075 Chr.10: 75028771-75028771 on Build GRCh38
rs755119518 Chr.10: 75028666-75028666 on Build GRCh38
rs773216336 Chr.10: 75028746-75028748 on Build GRCh38

View Full Product Details