Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22674785-22675058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2
Entrez Gene ID
2620
Gene Name
growth arrest specific 2
Gene Aliases
-
UniGene
Hs.711297
Gene Chromosome Location
Chr.11: 22626113-22885930 on Build GRCh38
Transcripts
NM_001143830.1 NM_005256.3 NM_177553.2 XM_011519971.2 XM_011519972.2 XM_011519975.1 XM_011519978.2 XM_017017528.1 XM_017017529.1 XM_017017530.1 XM_017017531.1 XM_017017532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838352 c.17G>A p.S6N Chr.11: 22674886-22674886 on Build GRCh38
4803248 c.62A>G p.Y21C Chr.11: 22674931-22674931 on Build GRCh38
5814354 c.106A>C p.M36L Chr.11: 22674975-22674975 on Build GRCh38
1353254 c.112G>A p.E38K Chr.11: 22674981-22674981 on Build GRCh38
275263 c.93T>G p.A31A Chr.11: 22674962-22674962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003064606 Chr.11: 22674925-22674925 on Build GRCh38
rs760677626 Chr.11: 22674899-22674899 on Build GRCh38
rs767695285 Chr.11: 22674897-22674897 on Build GRCh38
rs763324702 Chr.11: 22674864-22674864 on Build GRCh38
rs774918567 Chr.11: 22674972-22674972 on Build GRCh38
rs762936591 Chr.11: 22675019-22675019 on Build GRCh38
rs935439293 Chr.11: 22674856-22674856 on Build GRCh38
rs541251414 Chr.11: 22674975-22674975 on Build GRCh38
rs765165163 Chr.11: 22674900-22674900 on Build GRCh38
rs780736662 Chr.11: 22674833-22674833 on Build GRCh38
rs897558012 Chr.11: 22674915-22674915 on Build GRCh38
rs375761095 Chr.11: 22674955-22674955 on Build GRCh38
rs768130574 Chr.11: 22674824-22674824 on Build GRCh38
rs750350835 Chr.11: 22675018-22675018 on Build GRCh38
rs888035146 Chr.11: 22674903-22674903 on Build GRCh38
rs1047285480 Chr.11: 22674831-22674831 on Build GRCh38
rs754689232 Chr.11: 22674916-22674916 on Build GRCh38
rs780822556 Chr.11: 22674921-22674921 on Build GRCh38
rs752890533 Chr.11: 22674898-22674898 on Build GRCh38
rs747634423 Chr.11: 22674943-22674943 on Build GRCh38
rs772354095 Chr.11: 22674993-22674993 on Build GRCh38
rs751285443 Chr.11: 22675025-22675025 on Build GRCh38
rs1051078901 Chr.11: 22674874-22674874 on Build GRCh38
rs958760042 Chr.11: 22674959-22674959 on Build GRCh38
rs758180568 Chr.11: 22674907-22674907 on Build GRCh38
rs1038518247 Chr.11: 22674863-22674863 on Build GRCh38
rs764118419 Chr.11: 22675012-22675012 on Build GRCh38
rs866271246 Chr.11: 22674894-22674894 on Build GRCh38
rs542800103 Chr.11: 22674846-22674846 on Build GRCh38
rs146000909 Chr.11: 22674833-22674833 on Build GRCh38
rs749802933 Chr.11: 22674964-22674964 on Build GRCh38
rs760761357 Chr.11: 22675006-22675006 on Build GRCh38
rs759888461 Chr.11: 22674893-22674893 on Build GRCh38
rs1009559802 Chr.11: 22674982-22674982 on Build GRCh38
rs770266716 Chr.11: 22674860-22674860 on Build GRCh38
rs748596046 Chr.11: 22674843-22674843 on Build GRCh38
rs779873360 Chr.11: 22674909-22674909 on Build GRCh38
rs377124784 Chr.11: 22674818-22674818 on Build GRCh38
rs747651396 Chr.11: 22674996-22674996 on Build GRCh38
rs201780635 Chr.11: 22674960-22674960 on Build GRCh38
rs932190071 Chr.11: 22674871-22674871 on Build GRCh38
rs922571 Chr.11: 22674905-22674905 on Build GRCh38
rs147608463 Chr.11: 22674868-22674868 on Build GRCh38
rs372335985 Chr.11: 22674861-22674861 on Build GRCh38
rs751199626 Chr.11: 22674914-22674914 on Build GRCh38
rs766156870 Chr.11: 22675023-22675023 on Build GRCh38
rs775591142 Chr.11: 22674999-22674999 on Build GRCh38
rs1021489152 Chr.11: 22674995-22674995 on Build GRCh38
rs769207233 Chr.11: 22674950-22674950 on Build GRCh38
rs774687880 Chr.11: 22674892-22674892 on Build GRCh38
rs368894523 Chr.11: 22674965-22674965 on Build GRCh38
rs747812839 Chr.11: 22674836-22674836 on Build GRCh38

View Full Product Details