Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57625698-57625971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3502701 c.512C>T p.S171F Chr.15: 57625879-57625879 on Build GRCh38
4419216 c.504A>G p.A168A Chr.15: 57625871-57625871 on Build GRCh38
4402639 c.438G>A p.L146L Chr.15: 57625805-57625805 on Build GRCh38
344363 c.416C>T p.S139F Chr.15: 57625783-57625783 on Build GRCh38
4419217 c.411A>G p.A137A Chr.15: 57625871-57625871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779359863 Chr.15: 57625728-57625728 on Build GRCh38
rs1013835756 Chr.15: 57625725-57625725 on Build GRCh38
rs756342308 Chr.15: 57625776-57625776 on Build GRCh38
rs2069133 Chr.15: 57625871-57625871 on Build GRCh38
rs143011333 Chr.15: 57625882-57625882 on Build GRCh38
rs940647228 Chr.15: 57625817-57625817 on Build GRCh38
rs770945854 Chr.15: 57625907-57625907 on Build GRCh38
rs765033891 Chr.15: 57625762-57625762 on Build GRCh38
rs923154209 Chr.15: 57625858-57625858 on Build GRCh38
rs753815806 Chr.15: 57625787-57625787 on Build GRCh38
rs757816482 Chr.15: 57625796-57625796 on Build GRCh38
rs765347177 Chr.15: 57625870-57625870 on Build GRCh38
rs780745533 Chr.15: 57625886-57625886 on Build GRCh38
rs777925907 Chr.15: 57625777-57625777 on Build GRCh38
rs980901079 Chr.15: 57625734-57625734 on Build GRCh38
rs780392989 Chr.15: 57625823-57625823 on Build GRCh38
rs150919450 Chr.15: 57625746-57625746 on Build GRCh38
rs780409476 Chr.15: 57625877-57625877 on Build GRCh38
rs140782627 Chr.15: 57625767-57625767 on Build GRCh38
rs772484212 Chr.15: 57625804-57625804 on Build GRCh38
rs772396770 Chr.15: 57625729-57625729 on Build GRCh38
rs746353324 Chr.15: 57625801-57625801 on Build GRCh38
rs776777542 Chr.15: 57625925-57625925 on Build GRCh38
rs761821492 Chr.15: 57625754-57625754 on Build GRCh38
rs780470343 Chr.15: 57625834-57625834 on Build GRCh38
rs144926370 Chr.15: 57625843-57625843 on Build GRCh38
rs149704998 Chr.15: 57625921-57625921 on Build GRCh38
rs747323845 Chr.15: 57625878-57625878 on Build GRCh38
rs776224594 Chr.15: 57625828-57625828 on Build GRCh38
rs772785817 Chr.15: 57625898-57625898 on Build GRCh38
rs377700253 Chr.15: 57625844-57625844 on Build GRCh38
rs1013885059 Chr.15: 57625940-57625940 on Build GRCh38
rs377234100 Chr.15: 57625764-57625764 on Build GRCh38
rs747809908 Chr.15: 57625887-57625887 on Build GRCh38
rs200126298 Chr.15: 57625799-57625799 on Build GRCh38
rs774321796 Chr.15: 57625917-57625917 on Build GRCh38
rs1050243983 Chr.15: 57625889-57625889 on Build GRCh38
rs773068831 Chr.15: 57625832-57625832 on Build GRCh38
rs758836397 Chr.15: 57625873-57625873 on Build GRCh38
rs765236325 Chr.15: 57625932-57625932 on Build GRCh38
rs762090623 Chr.15: 57625929-57625929 on Build GRCh38
rs753039019 Chr.15: 57625773-57625773 on Build GRCh38
rs754061832 Chr.15: 57625851-57625851 on Build GRCh38
rs757331231 Chr.15: 57625853-57625853 on Build GRCh38
rs769313083 Chr.15: 57625831-57625831 on Build GRCh38
rs768222403 Chr.15: 57625743-57625743 on Build GRCh38
rs762492034 Chr.15: 57625829-57625829 on Build GRCh38
rs766250324 Chr.15: 57625768-57625768 on Build GRCh38
rs750443938 Chr.15: 57625941-57625941 on Build GRCh38
rs370946312 Chr.15: 57625842-57625842 on Build GRCh38
rs775172751 Chr.15: 57625735-57625735 on Build GRCh38
rs562945779 Chr.15: 57625924-57625924 on Build GRCh38
rs2958059 Chr.15: 57625892-57625892 on Build GRCh38
rs376537939 Chr.15: 57625824-57625824 on Build GRCh38

View Full Product Details