Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64719957-64720230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNS
Entrez Gene ID
2799
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
UniGene
Hs.334534
Gene Chromosome Location
Chr.12: 64713442-64759446 on Build GRCh38
Transcripts
NM_002076.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3734134 c.1555C>T p.R519C Chr.12: 64720047-64720047 on Build GRCh38
1363631 c.1516C>T p.R506W Chr.12: 64720086-64720086 on Build GRCh38
694794 c.1486G>C p.D496H Chr.12: 64720116-64720116 on Build GRCh38
1363630 c.1579G>A p.G527R Chr.12: 64720023-64720023 on Build GRCh38
7292946 c.1444A>G p.T482A Chr.12: 64720158-64720158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187312042 Chr.12: 64720046-64720046 on Build GRCh38
rs1000381159 Chr.12: 64720084-64720084 on Build GRCh38
rs1022346451 Chr.12: 64720017-64720017 on Build GRCh38
rs764819766 Chr.12: 64720189-64720189 on Build GRCh38
rs774658444 Chr.12: 64720138-64720138 on Build GRCh38
rs372487118 Chr.12: 64720067-64720067 on Build GRCh38
rs748361347 Chr.12: 64720096-64720096 on Build GRCh38
rs138790252 Chr.12: 64720149-64720149 on Build GRCh38
rs760143261 Chr.12: 64720145-64720145 on Build GRCh38
rs1006562871 Chr.12: 64720001-64720001 on Build GRCh38
rs141958901 Chr.12: 64720168-64720168 on Build GRCh38
rs980133508 Chr.12: 64720191-64720191 on Build GRCh38
rs753472052 Chr.12: 64720024-64720024 on Build GRCh38
rs781191329 Chr.12: 64720090-64720090 on Build GRCh38
rs767876998 Chr.12: 64720023-64720023 on Build GRCh38
rs376110509 Chr.12: 64719994-64719994 on Build GRCh38
rs369074040 Chr.12: 64720063-64720063 on Build GRCh38
rs373537057 Chr.12: 64720016-64720016 on Build GRCh38
rs749280926 Chr.12: 64720126-64720126 on Build GRCh38
rs767842253 Chr.12: 64720148-64720148 on Build GRCh38
rs768332556 Chr.12: 64720086-64720086 on Build GRCh38
rs773602473 Chr.12: 64720120-64720120 on Build GRCh38
rs904788730 Chr.12: 64720047-64720047 on Build GRCh38
rs770181525 Chr.12: 64720118-64720118 on Build GRCh38
rs756948381 Chr.12: 64720026-64720026 on Build GRCh38
rs749857419 Chr.12: 64720048-64720048 on Build GRCh38
rs192263369 Chr.12: 64720193-64720193 on Build GRCh38
rs765979939 Chr.12: 64720198-64720198 on Build GRCh38
rs757947342 Chr.12: 64720052-64720052 on Build GRCh38
rs771059884 Chr.12: 64720130-64720130 on Build GRCh38
rs746970479 Chr.12: 64720085-64720085 on Build GRCh38
rs889819546 Chr.12: 64720006-64720006 on Build GRCh38
rs34349183 Chr.12: 64720035-64720035 on Build GRCh38
rs757779410 Chr.12: 64720195-64720195 on Build GRCh38

View Full Product Details