Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39835617-39835890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTHD1
Entrez Gene ID
150350
Gene Name
ENTH domain containing 1
Gene Aliases
CACNA1I, dJ370M22.3
UniGene
Hs.474869
Gene Chromosome Location
Chr.22: 39743044-39893884 on Build GRCh38
Transcripts
NM_152512.3 XM_006724149.3 XM_006724150.3 XM_006724151.1 XM_006724153.3 XM_011529927.2 XM_011529928.2 XM_011529931.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632606 c.711G>A p.E237E Chr.22: 39835840-39835840 on Build GRCh38
299196 c.711+7C>A p.? Chr.22: 39835833-39835833 on Build GRCh38
6539654 c.690T>C p.I230I Chr.22: 39835861-39835861 on Build GRCh38
6095306 c.679C>A p.P227T Chr.22: 39835872-39835872 on Build GRCh38
355170 c.694G>T p.G232C Chr.22: 39835857-39835857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374320739 Chr.22: 39835806-39835806 on Build GRCh38
rs551924348 Chr.22: 39835720-39835720 on Build GRCh38
rs780323629 Chr.22: 39835831-39835831 on Build GRCh38
rs113756304 Chr.22: 39835655-39835655 on Build GRCh38
rs368531650 Chr.22: 39835800-39835800 on Build GRCh38
rs369569575 Chr.22: 39835798-39835798 on Build GRCh38
rs767484025 Chr.22: 39835791-39835791 on Build GRCh38
rs549444312 Chr.22: 39835645-39835645 on Build GRCh38
rs1032225228 Chr.22: 39835682-39835682 on Build GRCh38
rs116765273 Chr.22: 39835855-39835855 on Build GRCh38
rs566336218 Chr.22: 39835652-39835652 on Build GRCh38
rs112278629 Chr.22: 39835834-39835834 on Build GRCh38
rs910111446 Chr.22: 39835817-39835817 on Build GRCh38
rs781349511 Chr.22: 39835854-39835854 on Build GRCh38
rs750358124 Chr.22: 39835818-39835819 on Build GRCh38
rs1023738496 Chr.22: 39835757-39835757 on Build GRCh38
rs775299835 Chr.22: 39835839-39835839 on Build GRCh38
rs758447323 Chr.22: 39835803-39835803 on Build GRCh38
rs1038706344 Chr.22: 39835869-39835869 on Build GRCh38
rs999881952 Chr.22: 39835670-39835670 on Build GRCh38
rs982173033 Chr.22: 39835777-39835777 on Build GRCh38
rs770151688 Chr.22: 39835868-39835868 on Build GRCh38
rs956715712 Chr.22: 39835764-39835764 on Build GRCh38
rs917681967 Chr.22: 39835814-39835814 on Build GRCh38
rs1008671517 Chr.22: 39835644-39835644 on Build GRCh38
rs958024937 Chr.22: 39835708-39835708 on Build GRCh38
rs111734676 Chr.22: 39835779-39835779 on Build GRCh38
rs6001692 Chr.22: 39835709-39835709 on Build GRCh38
rs145258124 Chr.22: 39835653-39835653 on Build GRCh38
rs879856376 Chr.22: 39835678-39835678 on Build GRCh38
rs570081224 Chr.22: 39835766-39835766 on Build GRCh38

View Full Product Details