Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61810651-61810900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4 XM_011545022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390738 c.622C>T p.P208S Chr.11: 61810873-61810873 on Build GRCh38
7327355 c.615+2_616del94 p.? Chr.11: 61810879-61810972 on Build GRCh38
929528 c.624C>T p.P208P Chr.11: 61810871-61810871 on Build GRCh38
929527 c.714C>A p.F238L Chr.11: 61810781-61810781 on Build GRCh38
6657913 c.693C>T p.D231D Chr.11: 61810802-61810802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750558568 Chr.11: 61810699-61810699 on Build GRCh38
rs551159586 Chr.11: 61810774-61810774 on Build GRCh38
rs570924791 Chr.11: 61810791-61810791 on Build GRCh38
rs780543580 Chr.11: 61810748-61810748 on Build GRCh38
rs756269959 Chr.11: 61810700-61810700 on Build GRCh38
rs759817584 Chr.11: 61810878-61810878 on Build GRCh38
rs371880650 Chr.11: 61810747-61810747 on Build GRCh38
rs367574544 Chr.11: 61810775-61810775 on Build GRCh38
rs755346350 Chr.11: 61810737-61810737 on Build GRCh38
rs768557985 Chr.11: 61810778-61810778 on Build GRCh38
rs761814368 Chr.11: 61810823-61810823 on Build GRCh38
rs201926122 Chr.11: 61810870-61810870 on Build GRCh38
rs754132651 Chr.11: 61810736-61810736 on Build GRCh38
rs769293153 Chr.11: 61810756-61810756 on Build GRCh38
rs759067609 Chr.11: 61810739-61810748 on Build GRCh38
rs376564455 Chr.11: 61810754-61810754 on Build GRCh38
rs11548149 Chr.11: 61810875-61810875 on Build GRCh38
rs772099865 Chr.11: 61810824-61810824 on Build GRCh38
rs748648953 Chr.11: 61810746-61810746 on Build GRCh38
rs779303306 Chr.11: 61810740-61810740 on Build GRCh38
rs1028736610 Chr.11: 61810790-61810790 on Build GRCh38
rs1019520501 Chr.11: 61810678-61810678 on Build GRCh38
rs766451796 Chr.11: 61810871-61810871 on Build GRCh38
rs200077185 Chr.11: 61810705-61810705 on Build GRCh38
rs965304984 Chr.11: 61810712-61810712 on Build GRCh38
rs201323234 Chr.11: 61810847-61810847 on Build GRCh38

View Full Product Details