Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80653907-80654160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHFR
Entrez Gene ID
1719
Gene Name
dihydrofolate reductase
Gene Aliases
DHFRP1, DYR
UniGene
Hs.592364
Gene Chromosome Location
Chr.5: 80626226-80654981 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947752 c.98G>C p.R33T Chr.5: 80654046-80654046 on Build GRCh38
6326465 c.136+9G>A p.? Chr.5: 80653999-80653999 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044887788 Chr.5: 80653945-80653945 on Build GRCh38
rs572081813 Chr.5: 80654068-80654068 on Build GRCh38
rs778172287 Chr.5: 80654077-80654077 on Build GRCh38
rs764674335 Chr.5: 80654062-80654062 on Build GRCh38
rs773136060 Chr.5: 80654045-80654045 on Build GRCh38
rs767425256 Chr.5: 80654038-80654038 on Build GRCh38
rs201745474 Chr.5: 80654054-80654054 on Build GRCh38
rs944411304 Chr.5: 80653935-80653935 on Build GRCh38
rs769880102 Chr.5: 80653999-80653999 on Build GRCh38
rs1032250222 Chr.5: 80654011-80654011 on Build GRCh38
rs762459617 Chr.5: 80653998-80653998 on Build GRCh38
rs771048956 Chr.5: 80653968-80653968 on Build GRCh38
rs948539157 Chr.5: 80654108-80654108 on Build GRCh38
rs761687618 Chr.5: 80653981-80653982 on Build GRCh38
rs1019423802 Chr.5: 80654078-80654078 on Build GRCh38
rs965218942 Chr.5: 80654123-80654126 on Build GRCh38
rs1054100832 Chr.5: 80654085-80654085 on Build GRCh38
rs894323747 Chr.5: 80654096-80654096 on Build GRCh38
rs144847410 Chr.5: 80653988-80653988 on Build GRCh38
rs904499589 Chr.5: 80653967-80653967 on Build GRCh38
rs761785469 Chr.5: 80654025-80654025 on Build GRCh38
rs765924078 Chr.5: 80654055-80654055 on Build GRCh38
rs753426178 Chr.5: 80654060-80654060 on Build GRCh38
rs747303120 Chr.5: 80653964-80653964 on Build GRCh38
rs533184784 Chr.5: 80654134-80654134 on Build GRCh38
rs753212015 Chr.5: 80654064-80654064 on Build GRCh38
rs774452953 Chr.5: 80654021-80654021 on Build GRCh38
rs6151593 Chr.5: 80653984-80653984 on Build GRCh38
rs776823823 Chr.5: 80653981-80653981 on Build GRCh38
rs778049706 Chr.5: 80653958-80653958 on Build GRCh38
rs562836282 Chr.5: 80654129-80654129 on Build GRCh38
rs541144933 Chr.5: 80654117-80654117 on Build GRCh38
rs555346066 Chr.5: 80654061-80654061 on Build GRCh38

View Full Product Details