Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176499942-176500203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAF2
Entrez Gene ID
23197
Gene Name
Fas associated factor family member 2
Gene Aliases
ETEA, UBXD8, UBXN3B
UniGene
Hs.484242
Gene Chromosome Location
Chr.5: 176448333-176510074 on Build GRCh38
Transcripts
NM_014613.2 XM_011534475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614322 c.1051C>T p.P351S Chr.5: 176500042-176500042 on Build GRCh38
4422329 c.1119A>T p.R373R Chr.5: 176500110-176500110 on Build GRCh38
381215 c.1055C>T p.P352L Chr.5: 176500046-176500046 on Build GRCh38
1066027 c.1115C>A p.S372Y Chr.5: 176500106-176500106 on Build GRCh38
6170588 c.1118G>T p.R373L Chr.5: 176500109-176500109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767482024 Chr.5: 176500038-176500038 on Build GRCh38
rs764082385 Chr.5: 176500056-176500056 on Build GRCh38
rs376724942 Chr.5: 176500122-176500122 on Build GRCh38
rs760022482 Chr.5: 176500045-176500045 on Build GRCh38
rs569159857 Chr.5: 176500041-176500041 on Build GRCh38
rs572960612 Chr.5: 176500177-176500177 on Build GRCh38
rs879095365 Chr.5: 176500047-176500047 on Build GRCh38
rs748140249 Chr.5: 176500157-176500157 on Build GRCh38
rs922684538 Chr.5: 176500031-176500031 on Build GRCh38
rs749855390 Chr.5: 176500171-176500171 on Build GRCh38
rs941088892 Chr.5: 176500176-176500176 on Build GRCh38
rs574255369 Chr.5: 176500146-176500146 on Build GRCh38
rs767203421 Chr.5: 176500025-176500025 on Build GRCh38
rs908265642 Chr.5: 176500174-176500174 on Build GRCh38
rs774043842 Chr.5: 176499980-176499980 on Build GRCh38
rs935389411 Chr.5: 176500035-176500035 on Build GRCh38
rs759178977 Chr.5: 176499992-176499992 on Build GRCh38
rs976896396 Chr.5: 176499975-176499975 on Build GRCh38
rs147370701 Chr.5: 176500116-176500116 on Build GRCh38
rs778235421 Chr.5: 176500166-176500166 on Build GRCh38
rs767343185 Chr.5: 176500137-176500137 on Build GRCh38
rs781487779 Chr.5: 176500154-176500154 on Build GRCh38
rs755085571 Chr.5: 176500140-176500140 on Build GRCh38
rs752092758 Chr.5: 176500036-176500036 on Build GRCh38
rs554701483 Chr.5: 176500164-176500164 on Build GRCh38
rs753865513 Chr.5: 176500091-176500091 on Build GRCh38

View Full Product Details