Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1759527-1759752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSD
Entrez Gene ID
1509
Gene Name
cathepsin D
Gene Aliases
CLN10, CPSD, HEL-S-130P
UniGene
Hs.654447
Gene Chromosome Location
Chr.11: 1752752-1763992 on Build GRCh38
Transcripts
NM_001909.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4816898 c.320C>G p.S107C Chr.11: 1759548-1759548 on Build GRCh38
4031875 c.261G>A p.T87T Chr.11: 1759607-1759607 on Build GRCh38
6345238 c.306C>A p.N102K Chr.11: 1759562-1759562 on Build GRCh38
4392800 c.300C>T p.S100S Chr.11: 1759568-1759568 on Build GRCh38
4482762 c.263C>T p.P88L Chr.11: 1759605-1759605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368152533 Chr.11: 1759658-1759658 on Build GRCh38
rs199551387 Chr.11: 1759627-1759627 on Build GRCh38
rs754680487 Chr.11: 1759579-1759579 on Build GRCh38
rs542594251 Chr.11: 1759625-1759625 on Build GRCh38
rs372181453 Chr.11: 1759607-1759607 on Build GRCh38
rs375153172 Chr.11: 1759728-1759729 on Build GRCh38
rs531566910 Chr.11: 1759731-1759731 on Build GRCh38
rs11555042 Chr.11: 1759576-1759576 on Build GRCh38
rs996421254 Chr.11: 1759631-1759631 on Build GRCh38
rs796052400 Chr.11: 1759606-1759606 on Build GRCh38
rs111965770 Chr.11: 1759640-1759640 on Build GRCh38
rs767481255 Chr.11: 1759547-1759547 on Build GRCh38
rs927292240 Chr.11: 1759724-1759724 on Build GRCh38
rs374216275 Chr.11: 1759672-1759672 on Build GRCh38
rs957587676 Chr.11: 1759711-1759711 on Build GRCh38
rs547063584 Chr.11: 1759693-1759693 on Build GRCh38
rs753642334 Chr.11: 1759665-1759665 on Build GRCh38
rs752612332 Chr.11: 1759599-1759600 on Build GRCh38
rs966487833 Chr.11: 1759663-1759663 on Build GRCh38
rs767539066 Chr.11: 1759651-1759651 on Build GRCh38
rs796052407 Chr.11: 1759569-1759569 on Build GRCh38
rs1015068037 Chr.11: 1759709-1759709 on Build GRCh38
rs778616837 Chr.11: 1759668-1759668 on Build GRCh38
rs778220078 Chr.11: 1759580-1759580 on Build GRCh38
rs748897941 Chr.11: 1759575-1759575 on Build GRCh38
rs776022917 Chr.11: 1759620-1759620 on Build GRCh38
rs1042278213 Chr.11: 1759597-1759597 on Build GRCh38
rs748389548 Chr.11: 1759714-1759714 on Build GRCh38
rs112184520 Chr.11: 1759664-1759664 on Build GRCh38
rs2230067 Chr.11: 1759637-1759637 on Build GRCh38
rs981944847 Chr.11: 1759717-1759717 on Build GRCh38
rs771539723 Chr.11: 1759588-1759588 on Build GRCh38
rs897944478 Chr.11: 1759555-1759555 on Build GRCh38
rs1002089381 Chr.11: 1759586-1759586 on Build GRCh38
rs754798069 Chr.11: 1759667-1759667 on Build GRCh38
rs750415207 Chr.11: 1759561-1759561 on Build GRCh38
rs139154882 Chr.11: 1759601-1759601 on Build GRCh38
rs770281196 Chr.11: 1759710-1759710 on Build GRCh38
rs1049074086 Chr.11: 1759567-1759567 on Build GRCh38
rs770248571 Chr.11: 1759608-1759608 on Build GRCh38
rs141482597 Chr.11: 1759585-1759585 on Build GRCh38
rs532100424 Chr.11: 1759676-1759676 on Build GRCh38
rs993457414 Chr.11: 1759563-1759563 on Build GRCh38
rs750329344 Chr.11: 1759657-1759657 on Build GRCh38
rs587780915 Chr.11: 1759598-1759598 on Build GRCh38
rs758186555 Chr.11: 1759657-1759657 on Build GRCh38
rs142871900 Chr.11: 1759725-1759726 on Build GRCh38
rs369373285 Chr.11: 1759574-1759574 on Build GRCh38
rs549803615 Chr.11: 1759649-1759649 on Build GRCh38
rs371608763 Chr.11: 1759689-1759689 on Build GRCh38
rs145281838 Chr.11: 1759613-1759613 on Build GRCh38
rs944863011 Chr.11: 1759732-1759732 on Build GRCh38
rs748457876 Chr.11: 1759659-1759659 on Build GRCh38
rs147641822 Chr.11: 1759628-1759628 on Build GRCh38
rs924956136 Chr.11: 1759684-1759684 on Build GRCh38

View Full Product Details