Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46058468-46058699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG1
Entrez Gene ID
2565
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Gene Aliases
-
UniGene
Hs.375051
Gene Chromosome Location
Chr.4: 46035769-46124065 on Build GRCh38
Transcripts
NM_173536.3 XM_017007990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891807 c.659G>A p.W220* Chr.4: 46058589-46058589 on Build GRCh38
5466164 c.759C>T p.I253I Chr.4: 46058489-46058489 on Build GRCh38
5886340 c.699G>A p.W233* Chr.4: 46058549-46058549 on Build GRCh38
5427002 c.757A>C p.I253L Chr.4: 46058491-46058491 on Build GRCh38
354412 c.731G>A p.R244Q Chr.4: 46058517-46058517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762892106 Chr.4: 46058670-46058670 on Build GRCh38
rs780528950 Chr.4: 46058594-46058594 on Build GRCh38
rs746294744 Chr.4: 46058503-46058503 on Build GRCh38
rs181182185 Chr.4: 46058658-46058658 on Build GRCh38
rs752674369 Chr.4: 46058565-46058565 on Build GRCh38
rs773368605 Chr.4: 46058543-46058543 on Build GRCh38
rs1055865300 Chr.4: 46058633-46058633 on Build GRCh38
rs773359521 Chr.4: 46058653-46058653 on Build GRCh38
rs768183316 Chr.4: 46058663-46058664 on Build GRCh38
rs767426287 Chr.4: 46058525-46058525 on Build GRCh38
rs769081092 Chr.4: 46058640-46058640 on Build GRCh38
rs1007335424 Chr.4: 46058655-46058655 on Build GRCh38
rs267600166 Chr.4: 46058588-46058588 on Build GRCh38
rs200998867 Chr.4: 46058641-46058641 on Build GRCh38
rs745360102 Chr.4: 46058599-46058599 on Build GRCh38
rs749637208 Chr.4: 46058516-46058516 on Build GRCh38
rs775396111 Chr.4: 46058664-46058664 on Build GRCh38
rs756938824 Chr.4: 46058579-46058579 on Build GRCh38
rs139684526 Chr.4: 46058576-46058576 on Build GRCh38
rs766515295 Chr.4: 46058661-46058661 on Build GRCh38
rs772357681 Chr.4: 46058647-46058647 on Build GRCh38
rs373443309 Chr.4: 46058522-46058522 on Build GRCh38
rs765141807 Chr.4: 46058562-46058562 on Build GRCh38
rs142799145 Chr.4: 46058518-46058518 on Build GRCh38
rs1028522758 Chr.4: 46058600-46058600 on Build GRCh38
rs769209641 Chr.4: 46058517-46058517 on Build GRCh38
rs763936393 Chr.4: 46058575-46058575 on Build GRCh38
rs184497238 Chr.4: 46058607-46058607 on Build GRCh38
rs564336520 Chr.4: 46058507-46058507 on Build GRCh38
rs199600304 Chr.4: 46058635-46058635 on Build GRCh38
rs763811696 Chr.4: 46058672-46058672 on Build GRCh38
rs888456827 Chr.4: 46058549-46058549 on Build GRCh38
rs150607696 Chr.4: 46058554-46058554 on Build GRCh38
rs776187307 Chr.4: 46058509-46058509 on Build GRCh38
rs41310791 Chr.4: 46058498-46058498 on Build GRCh38
rs188859110 Chr.4: 46058646-46058646 on Build GRCh38
rs758188912 Chr.4: 46058567-46058567 on Build GRCh38
rs568991486 Chr.4: 46058615-46058615 on Build GRCh38
rs998346894 Chr.4: 46058589-46058589 on Build GRCh38
rs756392838 Chr.4: 46058500-46058500 on Build GRCh38
rs765192097 Chr.4: 46058630-46058631 on Build GRCh38

View Full Product Details