Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56398928-56399201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF1
Entrez Gene ID
1477
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
CstF-50, CstFp50
UniGene
Hs.172865
Gene Chromosome Location
Chr.20: 56392371-56404526 on Build GRCh38
Transcripts
NM_001033521.1 NM_001033522.1 NM_001324.2 XM_011528600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1412621 c.753delT p.F252fs*27 Chr.20: 56399074-56399074 on Build GRCh38
6637019 c.662G>A p.R221H Chr.20: 56398983-56398983 on Build GRCh38
1412620 c.703G>A p.G235R Chr.20: 56399024-56399024 on Build GRCh38
5885407 c.733G>A p.D245N Chr.20: 56399054-56399054 on Build GRCh38
4099642 c.813T>A p.N271K Chr.20: 56399134-56399134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781732308 Chr.20: 56399104-56399104 on Build GRCh38
rs139172063 Chr.20: 56398987-56398987 on Build GRCh38
rs747029931 Chr.20: 56399145-56399145 on Build GRCh38
rs140246355 Chr.20: 56399153-56399153 on Build GRCh38
rs761295702 Chr.20: 56399060-56399060 on Build GRCh38
rs771051331 Chr.20: 56399127-56399127 on Build GRCh38
rs879018304 Chr.20: 56399108-56399108 on Build GRCh38
rs568580084 Chr.20: 56398996-56398996 on Build GRCh38
rs149481390 Chr.20: 56398993-56398993 on Build GRCh38
rs755372580 Chr.20: 56398959-56398959 on Build GRCh38
rs367595219 Chr.20: 56399171-56399171 on Build GRCh38
rs766728394 Chr.20: 56399048-56399048 on Build GRCh38
rs147168971 Chr.20: 56399023-56399023 on Build GRCh38
rs764700659 Chr.20: 56399030-56399030 on Build GRCh38
rs201027648 Chr.20: 56399055-56399055 on Build GRCh38
rs6024851 Chr.20: 56398966-56398966 on Build GRCh38
rs781214099 Chr.20: 56398969-56398969 on Build GRCh38
rs771856540 Chr.20: 56398989-56398989 on Build GRCh38
rs374376023 Chr.20: 56399110-56399110 on Build GRCh38
rs926336294 Chr.20: 56399107-56399107 on Build GRCh38
rs756454676 Chr.20: 56399112-56399112 on Build GRCh38
rs757307761 Chr.20: 56399044-56399044 on Build GRCh38
rs775235261 Chr.20: 56399142-56399142 on Build GRCh38
rs748439069 Chr.20: 56398971-56398971 on Build GRCh38
rs768336341 Chr.20: 56399146-56399146 on Build GRCh38
rs377726257 Chr.20: 56399014-56399014 on Build GRCh38
rs984917876 Chr.20: 56398961-56398961 on Build GRCh38
rs376318288 Chr.20: 56398956-56398956 on Build GRCh38
rs752118196 Chr.20: 56399056-56399056 on Build GRCh38
rs769990325 Chr.20: 56398982-56398982 on Build GRCh38
rs568351281 Chr.20: 56399065-56399065 on Build GRCh38
rs371129354 Chr.20: 56399047-56399047 on Build GRCh38
rs778165698 Chr.20: 56399122-56399122 on Build GRCh38
rs762310241 Chr.20: 56399174-56399174 on Build GRCh38
rs749916815 Chr.20: 56399152-56399152 on Build GRCh38
rs763967458 Chr.20: 56398999-56398999 on Build GRCh38
rs773465704 Chr.20: 56398983-56398983 on Build GRCh38
rs373049003 Chr.20: 56399170-56399170 on Build GRCh38
rs749340913 Chr.20: 56399123-56399123 on Build GRCh38

View Full Product Details