Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 21570024-21570255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH11
Entrez Gene ID
8701
Gene Name
dynein axonemal heavy chain 11
Gene Aliases
CILD7, DNAHBL, DNAHC11, DNHBL, DPL11
UniGene
Hs.520245
Gene Chromosome Location
Chr.7: 21543215-21901568 on Build GRCh38
Transcripts
NM_001277115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4636502 c.1201G>T p.A401S Chr.7: 21570075-21570075 on Build GRCh38
7352419 c.1199C>T p.T400I Chr.7: 21570073-21570073 on Build GRCh38
5529895 c.1305C>T p.F435F Chr.7: 21570179-21570179 on Build GRCh38
239595 c.1307A>C p.N436T Chr.7: 21570181-21570181 on Build GRCh38
7364796 c.1337T>C p.M446T Chr.7: 21570211-21570211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528310220 Chr.7: 21570230-21570230 on Build GRCh38
rs376831401 Chr.7: 21570209-21570209 on Build GRCh38
rs765276431 Chr.7: 21570164-21570164 on Build GRCh38
rs900712695 Chr.7: 21570167-21570167 on Build GRCh38
rs1000957789 Chr.7: 21570114-21570114 on Build GRCh38
rs781498100 Chr.7: 21570077-21570077 on Build GRCh38
rs759050942 Chr.7: 21570163-21570163 on Build GRCh38
rs769216360 Chr.7: 21570105-21570105 on Build GRCh38
rs766041925 Chr.7: 21570155-21570155 on Build GRCh38
rs1029990395 Chr.7: 21570212-21570212 on Build GRCh38
rs751343231 Chr.7: 21570202-21570202 on Build GRCh38
rs886062172 Chr.7: 21570107-21570107 on Build GRCh38
rs780042783 Chr.7: 21570094-21570094 on Build GRCh38
rs72655982 Chr.7: 21570073-21570073 on Build GRCh38
rs1039701219 Chr.7: 21570165-21570165 on Build GRCh38
rs756345487 Chr.7: 21570216-21570216 on Build GRCh38
rs989108454 Chr.7: 21570197-21570197 on Build GRCh38
rs749372718 Chr.7: 21570232-21570232 on Build GRCh38
rs752632000 Chr.7: 21570166-21570166 on Build GRCh38
rs773158013 Chr.7: 21570138-21570138 on Build GRCh38
rs753357981 Chr.7: 21570160-21570160 on Build GRCh38
rs552249954 Chr.7: 21570106-21570106 on Build GRCh38
rs559143773 Chr.7: 21570185-21570185 on Build GRCh38
rs746037078 Chr.7: 21570081-21570081 on Build GRCh38
rs373862531 Chr.7: 21570132-21570132 on Build GRCh38
rs749371323 Chr.7: 21570101-21570101 on Build GRCh38
rs760295233 Chr.7: 21570149-21570149 on Build GRCh38
rs532486387 Chr.7: 21570174-21570175 on Build GRCh38
rs762191209 Chr.7: 21570131-21570131 on Build GRCh38
rs752546389 Chr.7: 21570057-21570057 on Build GRCh38
rs770023215 Chr.7: 21570085-21570085 on Build GRCh38

View Full Product Details