Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22209951-22210224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBLN1
Entrez Gene ID
340900
Gene Name
endogenous Bornavirus-like nucleoprotein 1
Gene Aliases
EBLN-1
UniGene
Hs.450519
Gene Chromosome Location
Chr.10: 22208814-22209983 on Build GRCh38
Transcripts
NM_001199938.1

SNP Detail

SNP ID Chromosome Location
rs962307447 Chr.10: 22210152-22210152 on Build GRCh38
rs540907515 Chr.10: 22210094-22210094 on Build GRCh38
rs976699411 Chr.10: 22210176-22210176 on Build GRCh38
rs77866090 Chr.10: 22210120-22210120 on Build GRCh38
rs955722114 Chr.10: 22210055-22210055 on Build GRCh38
rs908039031 Chr.10: 22210128-22210128 on Build GRCh38
rs965955035 Chr.10: 22210090-22210090 on Build GRCh38
rs1034820785 Chr.10: 22210102-22210102 on Build GRCh38
rs186562476 Chr.10: 22210113-22210113 on Build GRCh38
rs564271533 Chr.10: 22210106-22210106 on Build GRCh38
rs561315247 Chr.10: 22209979-22209979 on Build GRCh38
rs971765530 Chr.10: 22210063-22210063 on Build GRCh38
rs988897628 Chr.10: 22210057-22210057 on Build GRCh38
rs766816226 Chr.10: 22210068-22210068 on Build GRCh38
rs142227882 Chr.10: 22210091-22210091 on Build GRCh38
rs755142309 Chr.10: 22210186-22210186 on Build GRCh38
rs563500309 Chr.10: 22210132-22210132 on Build GRCh38
rs146971751 Chr.10: 22210021-22210021 on Build GRCh38
rs1010150039 Chr.10: 22209975-22209975 on Build GRCh38

View Full Product Details