Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52326251-52326480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH1
Entrez Gene ID
25981
Gene Name
dynein axonemal heavy chain 1
Gene Aliases
DNAHC1, HDHC7, HL-11, HL11, HSRF-1, XLHSRF-1
UniGene
Hs.655469
Gene Chromosome Location
Chr.3: 52313010-52400497 on Build GRCh38
Transcripts
NM_015512.4 XM_017006129.1 XM_017006130.1 XM_017006131.1 XM_017006132.1 XM_017006133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059854 c.561T>G p.P187P Chr.3: 52326294-52326294 on Build GRCh38
1284524 c.579G>T p.E193D Chr.3: 52326312-52326312 on Build GRCh38
3660642 c.576C>T p.I192I Chr.3: 52326309-52326309 on Build GRCh38
6379951 c.568A>G p.I190V Chr.3: 52326301-52326301 on Build GRCh38
6379950 c.568A>G p.I190V Chr.3: 52326301-52326301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761931981 Chr.3: 52326436-52326436 on Build GRCh38
rs762614713 Chr.3: 52326359-52326359 on Build GRCh38
rs768900414 Chr.3: 52326333-52326333 on Build GRCh38
rs756950796 Chr.3: 52326361-52326361 on Build GRCh38
rs780803009 Chr.3: 52326363-52326363 on Build GRCh38
rs1049398651 Chr.3: 52326440-52326440 on Build GRCh38
rs577717008 Chr.3: 52326437-52326437 on Build GRCh38
rs745750700 Chr.3: 52326321-52326321 on Build GRCh38
rs768066020 Chr.3: 52326282-52326282 on Build GRCh38
rs769831729 Chr.3: 52326347-52326348 on Build GRCh38
rs769746669 Chr.3: 52326324-52326324 on Build GRCh38
rs756704265 Chr.3: 52326287-52326287 on Build GRCh38
rs199966822 Chr.3: 52326346-52326346 on Build GRCh38
rs184549647 Chr.3: 52326338-52326338 on Build GRCh38
rs201444060 Chr.3: 52326284-52326284 on Build GRCh38
rs893263007 Chr.3: 52326445-52326445 on Build GRCh38
rs575963889 Chr.3: 52326370-52326371 on Build GRCh38
rs772256984 Chr.3: 52326345-52326345 on Build GRCh38
rs376123460 Chr.3: 52326318-52326318 on Build GRCh38
rs750251625 Chr.3: 52326364-52326364 on Build GRCh38
rs555661049 Chr.3: 52326309-52326309 on Build GRCh38
rs1040389896 Chr.3: 52326377-52326377 on Build GRCh38
rs188209063 Chr.3: 52326350-52326350 on Build GRCh38
rs1033283355 Chr.3: 52326439-52326439 on Build GRCh38
rs763376058 Chr.3: 52326352-52326353 on Build GRCh38
rs757070489 Chr.3: 52326310-52326310 on Build GRCh38
rs374684031 Chr.3: 52326365-52326365 on Build GRCh38
rs752219210 Chr.3: 52326353-52326353 on Build GRCh38
rs914030550 Chr.3: 52326288-52326288 on Build GRCh38
rs897630573 Chr.3: 52326374-52326374 on Build GRCh38
rs773711719 Chr.3: 52326351-52326352 on Build GRCh38
rs960771741 Chr.3: 52326280-52326280 on Build GRCh38
rs1033559716 Chr.3: 52326270-52326270 on Build GRCh38
rs774756098 Chr.3: 52326279-52326279 on Build GRCh38
rs763121110 Chr.3: 52326273-52326273 on Build GRCh38
rs181177631 Chr.3: 52326281-52326281 on Build GRCh38
rs61729450 Chr.3: 52326302-52326302 on Build GRCh38
rs993359793 Chr.3: 52326424-52326424 on Build GRCh38
rs761212989 Chr.3: 52326348-52326348 on Build GRCh38
rs35991561 Chr.3: 52326374-52326374 on Build GRCh38
rs201642506 Chr.3: 52326326-52326326 on Build GRCh38
rs368210345 Chr.3: 52326295-52326295 on Build GRCh38
rs996791826 Chr.3: 52326418-52326418 on Build GRCh38
rs369026320 Chr.3: 52326328-52326328 on Build GRCh38
rs200051758 Chr.3: 52326296-52326296 on Build GRCh38
rs759599544 Chr.3: 52326293-52326293 on Build GRCh38
rs919716008 Chr.3: 52326327-52326327 on Build GRCh38
rs900510879 Chr.3: 52326386-52326386 on Build GRCh38
rs764244843 Chr.3: 52326277-52326277 on Build GRCh38
rs575545601 Chr.3: 52326334-52326334 on Build GRCh38
rs540396977 Chr.3: 52326443-52326443 on Build GRCh38
rs762231833 Chr.3: 52326344-52326344 on Build GRCh38
rs751108875 Chr.3: 52326360-52326360 on Build GRCh38

View Full Product Details