Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168907460-168907733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6088461 c.691G>T p.D231Y Chr.2: 168907702-168907702 on Build GRCh38
148982 c.655G>C p.V219L Chr.2: 168907666-168907666 on Build GRCh38
337742 c.580G>C p.E194Q Chr.2: 168907591-168907591 on Build GRCh38
6668288 c.649T>C p.F217L Chr.2: 168907660-168907660 on Build GRCh38
4839521 c.693C>A p.D231E Chr.2: 168907704-168907704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761371281 Chr.2: 168907563-168907563 on Build GRCh38
rs748106178 Chr.2: 168907614-168907614 on Build GRCh38
rs550443332 Chr.2: 168907516-168907516 on Build GRCh38
rs779527018 Chr.2: 168907608-168907608 on Build GRCh38
rs973429156 Chr.2: 168907508-168907508 on Build GRCh38
rs779080955 Chr.2: 168907621-168907621 on Build GRCh38
rs141216504 Chr.2: 168907708-168907708 on Build GRCh38
rs754339473 Chr.2: 168907553-168907553 on Build GRCh38
rs745886276 Chr.2: 168907629-168907629 on Build GRCh38
rs749840850 Chr.2: 168907571-168907571 on Build GRCh38
rs755278632 Chr.2: 168907534-168907534 on Build GRCh38
rs758104751 Chr.2: 168907573-168907573 on Build GRCh38
rs749480279 Chr.2: 168907613-168907613 on Build GRCh38
rs35259259 Chr.2: 168907568-168907568 on Build GRCh38
rs200209268 Chr.2: 168907643-168907643 on Build GRCh38
rs183462054 Chr.2: 168907538-168907538 on Build GRCh38
rs973683318 Chr.2: 168907627-168907627 on Build GRCh38
rs139587795 Chr.2: 168907655-168907655 on Build GRCh38
rs767604486 Chr.2: 168907676-168907702 on Build GRCh38
rs764817338 Chr.2: 168907568-168907568 on Build GRCh38
rs492594 Chr.2: 168907666-168907666 on Build GRCh38
rs145217135 Chr.2: 168907700-168907700 on Build GRCh38
rs144077223 Chr.2: 168907710-168907710 on Build GRCh38
rs972971686 Chr.2: 168907702-168907702 on Build GRCh38
rs752499343 Chr.2: 168907694-168907694 on Build GRCh38
rs771117473 Chr.2: 168907517-168907517 on Build GRCh38
rs780384187 Chr.2: 168907713-168907713 on Build GRCh38
rs947868999 Chr.2: 168907598-168907598 on Build GRCh38
rs112063031 Chr.2: 168907633-168907633 on Build GRCh38
rs377582119 Chr.2: 168907610-168907610 on Build GRCh38
rs772144472 Chr.2: 168907539-168907539 on Build GRCh38
rs201094274 Chr.2: 168907620-168907620 on Build GRCh38
rs2232323 Chr.2: 168907631-168907631 on Build GRCh38
rs762351172 Chr.2: 168907656-168907656 on Build GRCh38
rs941581143 Chr.2: 168907501-168907501 on Build GRCh38
rs776103155 Chr.2: 168907647-168907647 on Build GRCh38
rs767421347 Chr.2: 168907690-168907690 on Build GRCh38
rs375527806 Chr.2: 168907675-168907675 on Build GRCh38
rs777548181 Chr.2: 168907707-168907707 on Build GRCh38
rs2232324 Chr.2: 168907638-168907638 on Build GRCh38
rs751241913 Chr.2: 168907676-168907676 on Build GRCh38
rs372931678 Chr.2: 168907552-168907552 on Build GRCh38
rs368489404 Chr.2: 168907549-168907549 on Build GRCh38
rs774232751 Chr.2: 168907530-168907530 on Build GRCh38
rs371639677 Chr.2: 168907697-168907697 on Build GRCh38
rs182708685 Chr.2: 168907693-168907693 on Build GRCh38
rs368644754 Chr.2: 168907677-168907677 on Build GRCh38
rs779851185 Chr.2: 168907632-168907632 on Build GRCh38
rs903728726 Chr.2: 168907611-168907611 on Build GRCh38

View Full Product Details