Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136206364-136206635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL1
Entrez Gene ID
2273
Gene Name
four and a half LIM domains 1
Gene Aliases
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
UniGene
Hs.435369
Gene Chromosome Location
Chr.X: 136146702-136211359 on Build GRCh38
Transcripts
NM_001159699.1 NM_001159700.1 NM_001159701.1 NM_001159702.2 NM_001159703.1 NM_001159704.1 NM_001167819.1 NM_001449.4 NR_027621.1 XM_006724743.2 XM_006724744.2 XM_006724745.3 XM_006724746.2 XM_006724747.2 XM_011531316.1 XM_017029357.1

SNP Detail

SNP ID Chromosome Location
rs886043917 Chr.X: 136206545-136206545 on Build GRCh38
rs776249397 Chr.X: 136206553-136206553 on Build GRCh38
rs770499208 Chr.X: 136206549-136206549 on Build GRCh38
rs772911535 Chr.X: 136206502-136206502 on Build GRCh38
rs368235882 Chr.X: 136206540-136206540 on Build GRCh38
rs113620195 Chr.X: 136206403-136206403 on Build GRCh38
rs182106777 Chr.X: 136206593-136206593 on Build GRCh38
rs146125558 Chr.X: 136206437-136206437 on Build GRCh38
rs781418161 Chr.X: 136206392-136206392 on Build GRCh38
rs759110550 Chr.X: 136206568-136206568 on Build GRCh38
rs111898308 Chr.X: 136206404-136206404 on Build GRCh38
rs374346567 Chr.X: 136206402-136206402 on Build GRCh38
rs11557265 Chr.X: 136206511-136206511 on Build GRCh38
rs780792982 Chr.X: 136206567-136206567 on Build GRCh38
rs754421860 Chr.X: 136206532-136206532 on Build GRCh38
rs769992006 Chr.X: 136206431-136206431 on Build GRCh38
rs111657067 Chr.X: 136206405-136206405 on Build GRCh38
rs763404867 Chr.X: 136206594-136206594 on Build GRCh38
rs140149764 Chr.X: 136206498-136206498 on Build GRCh38
rs111289709 Chr.X: 136206401-136206401 on Build GRCh38
rs764595951 Chr.X: 136206606-136206606 on Build GRCh38
rs372301312 Chr.X: 136206578-136206578 on Build GRCh38
rs11557264 Chr.X: 136206563-136206563 on Build GRCh38
rs748075349 Chr.X: 136206450-136206450 on Build GRCh38

View Full Product Details