Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162011762-162012033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP4
Entrez Gene ID
1803
Gene Name
dipeptidyl peptidase 4
Gene Aliases
ADABP, ADCP2, CD26, DPPIV, TP103
UniGene
Hs.368912
Gene Chromosome Location
Chr.2: 161992241-162074542 on Build GRCh38
Transcripts
NM_001935.3 XM_005246371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631378 c.1717A>G p.I573V Chr.2: 162011908-162011908 on Build GRCh38
5058183 c.1639T>A p.Y547N Chr.2: 162011986-162011986 on Build GRCh38
1008329 c.1731C>A p.S577R Chr.2: 162011894-162011894 on Build GRCh38
3568489 c.1745G>A p.G582E Chr.2: 162011880-162011880 on Build GRCh38
1008328 c.1753T>C p.Y585H Chr.2: 162011872-162011872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888572641 Chr.2: 162011896-162011896 on Build GRCh38
rs756171880 Chr.2: 162011793-162011793 on Build GRCh38
rs750167004 Chr.2: 162011925-162011925 on Build GRCh38
rs200732639 Chr.2: 162011928-162011928 on Build GRCh38
rs765516494 Chr.2: 162011927-162011927 on Build GRCh38
rs879478330 Chr.2: 162011888-162011888 on Build GRCh38
rs761296077 Chr.2: 162011861-162011861 on Build GRCh38
rs968979403 Chr.2: 162011799-162011799 on Build GRCh38
rs753980368 Chr.2: 162011989-162011989 on Build GRCh38
rs761446242 Chr.2: 162011903-162011903 on Build GRCh38
rs558709294 Chr.2: 162011858-162011858 on Build GRCh38
rs751419458 Chr.2: 162011951-162011951 on Build GRCh38
rs200106550 Chr.2: 162011856-162011856 on Build GRCh38
rs774794283 Chr.2: 162011809-162011809 on Build GRCh38
rs766293510 Chr.2: 162011931-162011931 on Build GRCh38
rs993006361 Chr.2: 162011990-162011990 on Build GRCh38
rs541319254 Chr.2: 162011975-162011975 on Build GRCh38
rs577060626 Chr.2: 162011868-162011868 on Build GRCh38
rs201576099 Chr.2: 162011832-162011832 on Build GRCh38
rs201691081 Chr.2: 162011952-162011952 on Build GRCh38
rs771336302 Chr.2: 162011807-162011807 on Build GRCh38
rs369643133 Chr.2: 162011914-162011914 on Build GRCh38
rs772588191 Chr.2: 162011841-162011841 on Build GRCh38
rs916894279 Chr.2: 162012005-162012005 on Build GRCh38
rs762689872 Chr.2: 162011926-162011926 on Build GRCh38
rs978894468 Chr.2: 162011845-162011845 on Build GRCh38
rs778038805 Chr.2: 162011800-162011800 on Build GRCh38
rs746219215 Chr.2: 162011829-162011829 on Build GRCh38
rs770800552 Chr.2: 162011880-162011880 on Build GRCh38
rs373721374 Chr.2: 162011957-162011957 on Build GRCh38
rs774080533 Chr.2: 162011993-162011993 on Build GRCh38
rs749517269 Chr.2: 162011804-162011804 on Build GRCh38
rs1007381077 Chr.2: 162011953-162011953 on Build GRCh38
rs748220838 Chr.2: 162011792-162011792 on Build GRCh38
rs1027492217 Chr.2: 162011881-162011881 on Build GRCh38

View Full Product Details