Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40310876-40311101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1B
Entrez Gene ID
10289
Gene Name
eukaryotic translation initiation factor 1B
Gene Aliases
GC20
UniGene
Hs.315230
Gene Chromosome Location
Chr.3: 40309682-40312424 on Build GRCh38
Transcripts
NM_005875.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680525 c.165_166insA p.K58fs*13 Chr.3: 40311026-40311027 on Build GRCh38
6097718 c.177T>A p.L59L Chr.3: 40311038-40311038 on Build GRCh38
5994780 c.176T>C p.L59P Chr.3: 40311037-40311037 on Build GRCh38
302121 c.73G>A p.A25T Chr.3: 40310934-40310934 on Build GRCh38
3775010 c.180G>A p.V60V Chr.3: 40311041-40311041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865939294 Chr.3: 40310922-40310922 on Build GRCh38
rs763097539 Chr.3: 40310990-40310990 on Build GRCh38
rs760588561 Chr.3: 40310959-40310959 on Build GRCh38
rs371724665 Chr.3: 40311014-40311014 on Build GRCh38
rs759772022 Chr.3: 40311067-40311067 on Build GRCh38
rs114327418 Chr.3: 40310966-40310966 on Build GRCh38
rs751708497 Chr.3: 40311062-40311062 on Build GRCh38
rs776682893 Chr.3: 40310972-40310972 on Build GRCh38
rs775081959 Chr.3: 40310933-40310933 on Build GRCh38
rs778562519 Chr.3: 40310927-40310927 on Build GRCh38

View Full Product Details