Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43530674-43530945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB6
Entrez Gene ID
64800
Gene Name
EF-hand calcium binding domain 6
Gene Aliases
DJBP, HSCBCIP1, dJ185D5.1
UniGene
Hs.658996
Gene Chromosome Location
Chr.22: 43528549-43812403 on Build GRCh38
Transcripts
NM_022785.3 NM_198856.2 XM_005261704.2 XM_005261705.3 XM_011530316.1 XM_011530317.2 XM_011530318.2 XM_011530319.2 XM_011530320.2 XM_011530321.2 XM_011530322.2 XM_011530323.2 XM_011530325.2 XM_011530327.2 XM_011530329.1 XM_017028910.1 XM_017028911.1 XM_017028912.1 NR_046563.1

Gene Symbol
EFCAB6-AS1
Entrez Gene ID
100874197
Gene Name
EFCAB6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.627997
Gene Chromosome Location
Chr.22: 43516254-43536605 on Build GRCh38
Transcripts
NM_022785.3 NM_198856.2 XM_005261704.2 XM_005261705.3 XM_011530316.1 XM_011530317.2 XM_011530318.2 XM_011530319.2 XM_011530320.2 XM_011530321.2 XM_011530322.2 XM_011530323.2 XM_011530325.2 XM_011530327.2 XM_011530329.1 XM_017028910.1 XM_017028911.1 XM_017028912.1 NR_046563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651933 c.4298T>C p.V1433A Chr.22: 43530900-43530900 on Build GRCh38
3390270 c.4365C>T p.S1455S Chr.22: 43530833-43530833 on Build GRCh38
3555136 c.4335C>T p.S1445S Chr.22: 43530863-43530863 on Build GRCh38
3555135 c.4372G>A p.D1458N Chr.22: 43530826-43530826 on Build GRCh38
726924 c.4321C>A p.R1441S Chr.22: 43530877-43530877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761597238 Chr.22: 43530859-43530859 on Build GRCh38
rs541720424 Chr.22: 43530799-43530799 on Build GRCh38
rs1006935458 Chr.22: 43530905-43530905 on Build GRCh38
rs781458260 Chr.22: 43530780-43530780 on Build GRCh38
rs763315688 Chr.22: 43530869-43530869 on Build GRCh38
rs963290884 Chr.22: 43530765-43530765 on Build GRCh38
rs889863960 Chr.22: 43530882-43530882 on Build GRCh38
rs767370813 Chr.22: 43530795-43530795 on Build GRCh38
rs751045331 Chr.22: 43530851-43530851 on Build GRCh38
rs374932960 Chr.22: 43530787-43530787 on Build GRCh38
rs750687549 Chr.22: 43530826-43530826 on Build GRCh38
rs751792593 Chr.22: 43530881-43530881 on Build GRCh38
rs771631087 Chr.22: 43530798-43530798 on Build GRCh38
rs778101008 Chr.22: 43530840-43530840 on Build GRCh38
rs201816198 Chr.22: 43530849-43530849 on Build GRCh38
rs144891068 Chr.22: 43530919-43530919 on Build GRCh38
rs151055081 Chr.22: 43530878-43530878 on Build GRCh38
rs768007652 Chr.22: 43530856-43530856 on Build GRCh38
rs770018138 Chr.22: 43530784-43530784 on Build GRCh38
rs141167337 Chr.22: 43530918-43530918 on Build GRCh38
rs141680193 Chr.22: 43530706-43530706 on Build GRCh38
rs769545422 Chr.22: 43530727-43530727 on Build GRCh38
rs1029579323 Chr.22: 43530848-43530848 on Build GRCh38
rs151282295 Chr.22: 43530835-43530835 on Build GRCh38
rs755092656 Chr.22: 43530883-43530883 on Build GRCh38
rs1018332079 Chr.22: 43530818-43530818 on Build GRCh38
rs761698490 Chr.22: 43530815-43530815 on Build GRCh38
rs201426447 Chr.22: 43530771-43530771 on Build GRCh38
rs750632684 Chr.22: 43530770-43530770 on Build GRCh38
rs745591218 Chr.22: 43530841-43530841 on Build GRCh38
rs539590253 Chr.22: 43530710-43530710 on Build GRCh38
rs144797247 Chr.22: 43530833-43530833 on Build GRCh38
rs776633265 Chr.22: 43530858-43530858 on Build GRCh38
rs781738975 Chr.22: 43530830-43530830 on Build GRCh38
rs563974290 Chr.22: 43530888-43530888 on Build GRCh38
rs992614985 Chr.22: 43530772-43530772 on Build GRCh38
rs915086992 Chr.22: 43530876-43530876 on Build GRCh38
rs935141947 Chr.22: 43530702-43530702 on Build GRCh38
rs149993265 Chr.22: 43530867-43530867 on Build GRCh38
rs140939056 Chr.22: 43530828-43530828 on Build GRCh38
rs746574690 Chr.22: 43530921-43530921 on Build GRCh38
rs760641608 Chr.22: 43530807-43530807 on Build GRCh38
rs373313660 Chr.22: 43530812-43530812 on Build GRCh38
rs769176388 Chr.22: 43530776-43530776 on Build GRCh38
rs1017559007 Chr.22: 43530764-43530764 on Build GRCh38
rs1045438907 Chr.22: 43530751-43530751 on Build GRCh38
rs773194460 Chr.22: 43530760-43530760 on Build GRCh38
rs933970627 Chr.22: 43530880-43530880 on Build GRCh38
rs980898350 Chr.22: 43530894-43530894 on Build GRCh38
rs764067760 Chr.22: 43530811-43530811 on Build GRCh38
rs751667777 Chr.22: 43530829-43530829 on Build GRCh38
rs771677982 Chr.22: 43530844-43530844 on Build GRCh38
rs774062183 Chr.22: 43530785-43530785 on Build GRCh38
rs756578495 Chr.22: 43530893-43530893 on Build GRCh38
rs201765619 Chr.22: 43530802-43530802 on Build GRCh38
rs753309592 Chr.22: 43530890-43530890 on Build GRCh38
rs556052088 Chr.22: 43530712-43530712 on Build GRCh38
rs778147732 Chr.22: 43530903-43530903 on Build GRCh38
rs926737361 Chr.22: 43530900-43530900 on Build GRCh38
rs200656934 Chr.22: 43530861-43530861 on Build GRCh38
rs762796463 Chr.22: 43530769-43530769 on Build GRCh38
rs372164113 Chr.22: 43530774-43530774 on Build GRCh38
rs529660338 Chr.22: 43530907-43530907 on Build GRCh38
rs764872032 Chr.22: 43530819-43530819 on Build GRCh38
rs35982593 Chr.22: 43530845-43530845 on Build GRCh38
rs796793248 Chr.22: 43530730-43530731 on Build GRCh38

View Full Product Details