Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32271713-32271962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPC1
Entrez Gene ID
80314
Gene Name
enhancer of polycomb homolog 1
Gene Aliases
Epl1
UniGene
Hs.167805
Gene Chromosome Location
Chr.10: 32267716-32378798 on Build GRCh38
Transcripts
NM_001272004.1 NM_001272019.2 NM_001282391.1 NM_025209.3 XM_006717510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654803 c.2267G>C p.R756P Chr.10: 32271725-32271725 on Build GRCh38
4411410 c.2204T>A p.L735Q Chr.10: 32271788-32271788 on Build GRCh38
1284752 c.2088C>T p.G696G Chr.10: 32271904-32271904 on Build GRCh38
6517785 c.2244G>T p.Q748H Chr.10: 32271748-32271748 on Build GRCh38
3985592 c.2127T>C p.H709H Chr.10: 32271865-32271865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777409070 Chr.10: 32271843-32271843 on Build GRCh38
rs114249238 Chr.10: 32271822-32271822 on Build GRCh38
rs751003082 Chr.10: 32271740-32271740 on Build GRCh38
rs770907136 Chr.10: 32271769-32271769 on Build GRCh38
rs200728281 Chr.10: 32271927-32271927 on Build GRCh38
rs748927703 Chr.10: 32271768-32271768 on Build GRCh38
rs775625085 Chr.10: 32271928-32271931 on Build GRCh38
rs200028097 Chr.10: 32271844-32271844 on Build GRCh38
rs755733746 Chr.10: 32271842-32271842 on Build GRCh38
rs759309767 Chr.10: 32271799-32271799 on Build GRCh38
rs192787659 Chr.10: 32271878-32271878 on Build GRCh38
rs771864906 Chr.10: 32271852-32271852 on Build GRCh38
rs761820723 Chr.10: 32271921-32271921 on Build GRCh38
rs144599692 Chr.10: 32271884-32271884 on Build GRCh38
rs768166993 Chr.10: 32271828-32271828 on Build GRCh38
rs903854727 Chr.10: 32271906-32271906 on Build GRCh38
rs762014881 Chr.10: 32271832-32271832 on Build GRCh38
rs1037274256 Chr.10: 32271742-32271742 on Build GRCh38
rs747631521 Chr.10: 32271753-32271753 on Build GRCh38
rs764131261 Chr.10: 32271827-32271827 on Build GRCh38
rs964815987 Chr.10: 32271913-32271913 on Build GRCh38
rs758739493 Chr.10: 32271838-32271838 on Build GRCh38
rs776693583 Chr.10: 32271909-32271909 on Build GRCh38
rs780455340 Chr.10: 32271839-32271839 on Build GRCh38
rs878879008 Chr.10: 32271892-32271892 on Build GRCh38
rs774013896 Chr.10: 32271795-32271795 on Build GRCh38
rs150041891 Chr.10: 32271823-32271823 on Build GRCh38
rs765430708 Chr.10: 32271835-32271835 on Build GRCh38
rs777608475 Chr.10: 32271764-32271764 on Build GRCh38
rs769387859 Chr.10: 32271761-32271761 on Build GRCh38
rs754114351 Chr.10: 32271829-32271829 on Build GRCh38
rs760588461 Chr.10: 32271885-32271885 on Build GRCh38
rs758830646 Chr.10: 32271747-32271747 on Build GRCh38
rs988030234 Chr.10: 32271862-32271862 on Build GRCh38
rs752082009 Chr.10: 32271840-32271840 on Build GRCh38
rs768787071 Chr.10: 32271901-32271901 on Build GRCh38
rs765334182 Chr.10: 32271883-32271888 on Build GRCh38
rs145765564 Chr.10: 32271850-32271850 on Build GRCh38
rs138540076 Chr.10: 32271851-32271851 on Build GRCh38
rs770496925 Chr.10: 32271845-32271845 on Build GRCh38
rs868231255 Chr.10: 32271841-32271844 on Build GRCh38
rs372077598 Chr.10: 32271802-32271802 on Build GRCh38
rs750764581 Chr.10: 32271836-32271836 on Build GRCh38
rs867993822 Chr.10: 32271836-32271844 on Build GRCh38
rs759560121 Chr.10: 32271877-32271882 on Build GRCh38
rs780551835 Chr.10: 32271749-32271749 on Build GRCh38

View Full Product Details