Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108900331-108900604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAO
Entrez Gene ID
1610
Gene Name
D-amino acid oxidase
Gene Aliases
DAAO, DAMOX, OXDA
UniGene
Hs.113227
Gene Chromosome Location
Chr.12: 108880073-108901043 on Build GRCh38
Transcripts
NM_001917.4 XM_005268692.3 XM_011538004.2 XM_011538005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934659 c.995G>T p.R332I Chr.12: 108900486-108900486 on Build GRCh38
5773887 c.1038C>T p.H346H Chr.12: 108900529-108900529 on Build GRCh38
6888756 c.1003G>A p.E335K Chr.12: 108900494-108900494 on Build GRCh38
4038441 c.1010A>C p.K337T Chr.12: 108900501-108900501 on Build GRCh38
3967960 c.969C>T p.A323A Chr.12: 108900460-108900460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781208737 Chr.12: 108900423-108900423 on Build GRCh38
rs185053494 Chr.12: 108900385-108900385 on Build GRCh38
rs754828923 Chr.12: 108900418-108900418 on Build GRCh38
rs190601584 Chr.12: 108900513-108900513 on Build GRCh38
rs771193408 Chr.12: 108900530-108900530 on Build GRCh38
rs200970049 Chr.12: 108900474-108900474 on Build GRCh38
rs376435571 Chr.12: 108900510-108900510 on Build GRCh38
rs751062244 Chr.12: 108900487-108900487 on Build GRCh38
rs201879447 Chr.12: 108900519-108900519 on Build GRCh38
rs202101905 Chr.12: 108900410-108900410 on Build GRCh38
rs200028465 Chr.12: 108900460-108900460 on Build GRCh38
rs553140906 Chr.12: 108900557-108900557 on Build GRCh38
rs761222054 Chr.12: 108900374-108900374 on Build GRCh38
rs768785859 Chr.12: 108900554-108900554 on Build GRCh38
rs777596901 Chr.12: 108900507-108900507 on Build GRCh38
rs4262766 Chr.12: 108900483-108900483 on Build GRCh38
rs770250800 Chr.12: 108900566-108900566 on Build GRCh38
rs768996759 Chr.12: 108900449-108900449 on Build GRCh38
rs757967086 Chr.12: 108900388-108900388 on Build GRCh38
rs770340579 Chr.12: 108900458-108900458 on Build GRCh38
rs764489974 Chr.12: 108900377-108900377 on Build GRCh38
rs374840450 Chr.12: 108900573-108900573 on Build GRCh38
rs746150850 Chr.12: 108900431-108900431 on Build GRCh38
rs747691375 Chr.12: 108900383-108900389 on Build GRCh38
rs762262174 Chr.12: 108900383-108900383 on Build GRCh38
rs774054039 Chr.12: 108900578-108900578 on Build GRCh38
rs531980044 Chr.12: 108900568-108900568 on Build GRCh38
rs866687761 Chr.12: 108900380-108900380 on Build GRCh38
rs575170073 Chr.12: 108900434-108900434 on Build GRCh38
rs3741773 Chr.12: 108900562-108900562 on Build GRCh38
rs200091028 Chr.12: 108900386-108900386 on Build GRCh38
rs191345652 Chr.12: 108900371-108900371 on Build GRCh38
rs143732132 Chr.12: 108900525-108900525 on Build GRCh38
rs201607867 Chr.12: 108900381-108900381 on Build GRCh38
rs564918916 Chr.12: 108900538-108900538 on Build GRCh38
rs199548432 Chr.12: 108900516-108900516 on Build GRCh38
rs747146248 Chr.12: 108900444-108900444 on Build GRCh38
rs773114925 Chr.12: 108900556-108900557 on Build GRCh38
rs745885794 Chr.12: 108900360-108900360 on Build GRCh38
rs759096607 Chr.12: 108900494-108900494 on Build GRCh38
rs747837081 Chr.12: 108900425-108900425 on Build GRCh38
rs925424619 Chr.12: 108900522-108900522 on Build GRCh38
rs867674559 Chr.12: 108900403-108900403 on Build GRCh38
rs563868242 Chr.12: 108900496-108900496 on Build GRCh38
rs755937780 Chr.12: 108900430-108900430 on Build GRCh38
rs762733390 Chr.12: 108900476-108900476 on Build GRCh38
rs770647158 Chr.12: 108900577-108900577 on Build GRCh38
rs200600877 Chr.12: 108900370-108900370 on Build GRCh38
rs772119645 Chr.12: 108900433-108900433 on Build GRCh38
rs781537569 Chr.12: 108900558-108900558 on Build GRCh38
rs1039685563 Chr.12: 108900561-108900561 on Build GRCh38
rs762017824 Chr.12: 108900457-108900457 on Build GRCh38
rs777208721 Chr.12: 108900452-108900452 on Build GRCh38
rs201376696 Chr.12: 108900399-108900399 on Build GRCh38
rs752743261 Chr.12: 108900505-108900505 on Build GRCh38

View Full Product Details