Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76265509-76265754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBALD2
Entrez Gene ID
283991
Gene Name
UBA like domain containing 2
Gene Aliases
FAM100B
UniGene
Hs.744981
Gene Chromosome Location
Chr.17: 76265205-76271298 on Build GRCh38
Transcripts
NM_182565.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4560604 c.84G>T p.A28A Chr.17: 76265589-76265589 on Build GRCh38
3522513 c.33G>A p.Q11Q Chr.17: 76265538-76265538 on Build GRCh38
3668060 c.12C>T p.N4N Chr.17: 76265517-76265517 on Build GRCh38
707151 c.57G>A p.L19L Chr.17: 76265562-76265562 on Build GRCh38
3749416 c.91T>C p.L31L Chr.17: 76265596-76265596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989859637 Chr.17: 76265680-76265680 on Build GRCh38
rs746575564 Chr.17: 76265628-76265628 on Build GRCh38
rs768300658 Chr.17: 76265629-76265629 on Build GRCh38
rs540289631 Chr.17: 76265730-76265730 on Build GRCh38
rs913921821 Chr.17: 76265688-76265688 on Build GRCh38
rs945414457 Chr.17: 76265693-76265693 on Build GRCh38
rs764635903 Chr.17: 76265563-76265563 on Build GRCh38
rs143586386 Chr.17: 76265568-76265568 on Build GRCh38
rs773120450 Chr.17: 76265650-76265650 on Build GRCh38
rs199683901 Chr.17: 76265580-76265580 on Build GRCh38
rs752892377 Chr.17: 76265532-76265532 on Build GRCh38
rs577330168 Chr.17: 76265649-76265649 on Build GRCh38
rs776236322 Chr.17: 76265636-76265636 on Build GRCh38
rs2585751 Chr.17: 76265596-76265596 on Build GRCh38
rs746204635 Chr.17: 76265588-76265588 on Build GRCh38
rs112233717 Chr.17: 76265735-76265735 on Build GRCh38
rs756118569 Chr.17: 76265549-76265549 on Build GRCh38
rs371046028 Chr.17: 76265537-76265537 on Build GRCh38
rs748104293 Chr.17: 76265639-76265639 on Build GRCh38

View Full Product Details