Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 198167781-198168008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM157A
Entrez Gene ID
728262
Gene Name
family with sequence similarity 157 member A
Gene Aliases
-
UniGene
Hs.744675
Gene Chromosome Location
Chr.3: 198152155-198198958 on Build GRCh38
Transcripts
NM_001145248.1 XM_017007125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595865 c.1036G>A p.A346T Chr.3: 198167823-198167823 on Build GRCh38
3774099 c.1055A>G p.E352G Chr.3: 198167842-198167842 on Build GRCh38
5795185 c.1032T>C p.A344A Chr.3: 198167819-198167819 on Build GRCh38
5034209 c.1035A>G p.S345S Chr.3: 198167822-198167822 on Build GRCh38
227730 c.1010G>A p.R337K Chr.3: 198167797-198167797 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766303947 Chr.3: 198167873-198167873 on Build GRCh38
rs951699443 Chr.3: 198167899-198167899 on Build GRCh38
rs61793577 Chr.3: 198167929-198167929 on Build GRCh38
rs556321333 Chr.3: 198167965-198167965 on Build GRCh38
rs752942806 Chr.3: 198167884-198167884 on Build GRCh38
rs762841595 Chr.3: 198167872-198167872 on Build GRCh38
rs544687171 Chr.3: 198167964-198167964 on Build GRCh38
rs369839941 Chr.3: 198167896-198167896 on Build GRCh38
rs61793576 Chr.3: 198167842-198167842 on Build GRCh38
rs375813333 Chr.3: 198167855-198167855 on Build GRCh38
rs995755743 Chr.3: 198167836-198167836 on Build GRCh38
rs574759702 Chr.3: 198167966-198167966 on Build GRCh38
rs187418814 Chr.3: 198167910-198167910 on Build GRCh38
rs542186252 Chr.3: 198167980-198167980 on Build GRCh38
rs576897057 Chr.3: 198167901-198167901 on Build GRCh38
rs772598469 Chr.3: 198167819-198167819 on Build GRCh38
rs367693829 Chr.3: 198167922-198167922 on Build GRCh38
rs539610541 Chr.3: 198167886-198167886 on Build GRCh38
rs61793575 Chr.3: 198167823-198167823 on Build GRCh38
rs554014939 Chr.3: 198167813-198167813 on Build GRCh38
rs961963894 Chr.3: 198167812-198167812 on Build GRCh38
rs577363293 Chr.3: 198167945-198167945 on Build GRCh38
rs867109846 Chr.3: 198167927-198167927 on Build GRCh38
rs768875295 Chr.3: 198167841-198167841 on Build GRCh38
rs762015189 Chr.3: 198167848-198167848 on Build GRCh38
rs143213060 Chr.3: 198167849-198167849 on Build GRCh38
rs775462675 Chr.3: 198167822-198167822 on Build GRCh38

View Full Product Details