Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62567334-62567599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1G
Entrez Gene ID
1937
Gene Name
eukaryotic translation elongation factor 1 gamma
Gene Aliases
EF1G, GIG35
UniGene
Hs.144835
Gene Chromosome Location
Chr.11: 62559601-62573988 on Build GRCh38
Transcripts
NM_001404.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6055021 c.565C>T p.R189C Chr.11: 62567486-62567486 on Build GRCh38
4400713 c.596A>G p.Q199R Chr.11: 62567455-62567455 on Build GRCh38
6859768 c.691C>T p.R231C Chr.11: 62567510-62567510 on Build GRCh38
165349 c.633G>A p.E211E Chr.11: 62567418-62567418 on Build GRCh38
3986300 c.611T>G p.L204W Chr.11: 62567440-62567440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775420753 Chr.11: 62567369-62567369 on Build GRCh38
rs769597769 Chr.11: 62567451-62567451 on Build GRCh38
rs761084418 Chr.11: 62567520-62567520 on Build GRCh38
rs1025706038 Chr.11: 62567427-62567427 on Build GRCh38
rs745605867 Chr.11: 62567450-62567450 on Build GRCh38
rs762212809 Chr.11: 62567385-62567387 on Build GRCh38
rs571268030 Chr.11: 62567499-62567499 on Build GRCh38
rs751212230 Chr.11: 62567441-62567441 on Build GRCh38
rs749263175 Chr.11: 62567484-62567484 on Build GRCh38
rs766947119 Chr.11: 62567412-62567412 on Build GRCh38
rs946252182 Chr.11: 62567559-62567559 on Build GRCh38
rs376662848 Chr.11: 62567570-62567570 on Build GRCh38
rs549342142 Chr.11: 62567374-62567374 on Build GRCh38
rs374064017 Chr.11: 62567384-62567384 on Build GRCh38
rs768757371 Chr.11: 62567492-62567492 on Build GRCh38
rs754379472 Chr.11: 62567561-62567561 on Build GRCh38
rs1061093 Chr.11: 62567436-62567436 on Build GRCh38
rs962584710 Chr.11: 62567388-62567388 on Build GRCh38
rs945173082 Chr.11: 62567571-62567571 on Build GRCh38
rs761989563 Chr.11: 62567396-62567396 on Build GRCh38
rs756548804 Chr.11: 62567407-62567407 on Build GRCh38
rs750712436 Chr.11: 62567552-62567557 on Build GRCh38
rs370728833 Chr.11: 62567543-62567543 on Build GRCh38
rs151279428 Chr.11: 62567475-62567475 on Build GRCh38
rs762969760 Chr.11: 62567373-62567373 on Build GRCh38
rs77818737 Chr.11: 62567422-62567422 on Build GRCh38
rs750880029 Chr.11: 62567402-62567402 on Build GRCh38
rs756927362 Chr.11: 62567445-62567445 on Build GRCh38
rs773579698 Chr.11: 62567504-62567504 on Build GRCh38
rs377344394 Chr.11: 62567503-62567503 on Build GRCh38
rs780903665 Chr.11: 62567449-62567449 on Build GRCh38
rs754430580 Chr.11: 62567435-62567435 on Build GRCh38
rs377238340 Chr.11: 62567400-62567400 on Build GRCh38
rs767875798 Chr.11: 62567469-62567471 on Build GRCh38
rs774349008 Chr.11: 62567496-62567496 on Build GRCh38

View Full Product Details