Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45350484-45350713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC2
Entrez Gene ID
2068
Gene Name
ERCC excision repair 2, TFIIH core complex helicase subunit
Gene Aliases
COFS2, EM9, TFIIH, TTD, TTD1, XPD
UniGene
Hs.487294
Gene Chromosome Location
Chr.19: 45349837-45370647 on Build GRCh38
Transcripts
XM_011526611.2 XM_017026467.1 NM_177417.2

Gene Symbol
KLC3
Entrez Gene ID
147700
Gene Name
kinesin light chain 3
Gene Aliases
KLC2, KLC2L, KLCt, KNS2B
UniGene
Hs.298079
Gene Chromosome Location
Chr.19: 45340740-45351520 on Build GRCh38
Transcripts
XM_011526611.2 XM_017026467.1 NM_177417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6696170 c.1242C>A p.P414P Chr.19: 45350521-45350521 on Build GRCh38
6696169 c.1238C>T p.A413V Chr.19: 45350517-45350517 on Build GRCh38
5452115 c.1250G>A p.G417D Chr.19: 45350529-45350529 on Build GRCh38
5914096 c.1235-7C>T p.? Chr.19: 45350507-45350507 on Build GRCh38
4928495 c.1311G>A p.E437E Chr.19: 45350679-45350679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758698292 Chr.19: 45350570-45350570 on Build GRCh38
rs748292230 Chr.19: 45350589-45350606 on Build GRCh38
rs750327046 Chr.19: 45350522-45350522 on Build GRCh38
rs368890796 Chr.19: 45350657-45350657 on Build GRCh38
rs773980237 Chr.19: 45350635-45350636 on Build GRCh38
rs767434756 Chr.19: 45350672-45350672 on Build GRCh38
rs770808255 Chr.19: 45350508-45350508 on Build GRCh38
rs778628117 Chr.19: 45350646-45350646 on Build GRCh38
rs190169301 Chr.19: 45350542-45350542 on Build GRCh38
rs923086524 Chr.19: 45350516-45350516 on Build GRCh38
rs764543395 Chr.19: 45350544-45350544 on Build GRCh38
rs779895784 Chr.19: 45350686-45350686 on Build GRCh38
rs776994753 Chr.19: 45350611-45350611 on Build GRCh38
rs768264781 Chr.19: 45350604-45350604 on Build GRCh38
rs757227601 Chr.19: 45350640-45350640 on Build GRCh38
rs896034965 Chr.19: 45350687-45350687 on Build GRCh38
rs763385091 Chr.19: 45350664-45350664 on Build GRCh38
rs774943834 Chr.19: 45350599-45350599 on Build GRCh38
rs199738278 Chr.19: 45350543-45350543 on Build GRCh38
rs765131448 Chr.19: 45350676-45350676 on Build GRCh38
rs371191877 Chr.19: 45350572-45350572 on Build GRCh38
rs756994856 Chr.19: 45350585-45350585 on Build GRCh38
rs763219085 Chr.19: 45350626-45350626 on Build GRCh38
rs771479406 Chr.19: 45350665-45350665 on Build GRCh38
rs371863089 Chr.19: 45350628-45350628 on Build GRCh38
rs749002991 Chr.19: 45350506-45350506 on Build GRCh38
rs766909609 Chr.19: 45350668-45350682 on Build GRCh38
rs780520784 Chr.19: 45350691-45350691 on Build GRCh38
rs369131208 Chr.19: 45350689-45350689 on Build GRCh38
rs983882094 Chr.19: 45350563-45350563 on Build GRCh38
rs771347899 Chr.19: 45350628-45350639 on Build GRCh38
rs570908802 Chr.19: 45350511-45350511 on Build GRCh38
rs368464700 Chr.19: 45350565-45350565 on Build GRCh38
rs747451288 Chr.19: 45350621-45350643 on Build GRCh38
rs374863110 Chr.19: 45350601-45350601 on Build GRCh38
rs749304819 Chr.19: 45350581-45350581 on Build GRCh38
rs376739020 Chr.19: 45350651-45350651 on Build GRCh38
rs773809752 Chr.19: 45350599-45350616 on Build GRCh38
rs757200055 Chr.19: 45350511-45350511 on Build GRCh38
rs560356289 Chr.19: 45350669-45350669 on Build GRCh38
rs770358528 Chr.19: 45350662-45350662 on Build GRCh38
rs758179169 Chr.19: 45350683-45350683 on Build GRCh38
rs777237318 Chr.19: 45350554-45350554 on Build GRCh38
rs373353596 Chr.19: 45350650-45350650 on Build GRCh38
rs368179138 Chr.19: 45350613-45350613 on Build GRCh38
rs762078454 Chr.19: 45350627-45350627 on Build GRCh38
rs755958597 Chr.19: 45350679-45350680 on Build GRCh38
rs777548567 Chr.19: 45350580-45350580 on Build GRCh38
rs375060034 Chr.19: 45350675-45350675 on Build GRCh38
rs771500038 Chr.19: 45350530-45350530 on Build GRCh38
rs755181966 Chr.19: 45350583-45350599 on Build GRCh38
rs375243876 Chr.19: 45350637-45350637 on Build GRCh38
rs776818817 Chr.19: 45350629-45350630 on Build GRCh38
rs750671148 Chr.19: 45350561-45350561 on Build GRCh38
rs529405107 Chr.19: 45350670-45350670 on Build GRCh38
rs369584425 Chr.19: 45350648-45350648 on Build GRCh38
rs1043613289 Chr.19: 45350583-45350583 on Build GRCh38
rs748545288 Chr.19: 45350661-45350661 on Build GRCh38
rs373548688 Chr.19: 45350693-45350693 on Build GRCh38
rs772036328 Chr.19: 45350590-45350607 on Build GRCh38
rs528482467 Chr.19: 45350662-45350663 on Build GRCh38
rs779694964 Chr.19: 45350571-45350572 on Build GRCh38
rs763680736 Chr.19: 45350634-45350635 on Build GRCh38
rs1040897551 Chr.19: 45350666-45350666 on Build GRCh38
rs762066937 Chr.19: 45350556-45350556 on Build GRCh38
rs779144746 Chr.19: 45350588-45350589 on Build GRCh38
rs749546141 Chr.19: 45350578-45350588 on Build GRCh38
rs745583423 Chr.19: 45350526-45350526 on Build GRCh38
rs367610871 Chr.19: 45350588-45350588 on Build GRCh38
rs765453943 Chr.19: 45350620-45350620 on Build GRCh38
rs754674282 Chr.19: 45350690-45350690 on Build GRCh38
rs994103837 Chr.19: 45350677-45350677 on Build GRCh38
rs773673888 Chr.19: 45350663-45350663 on Build GRCh38
rs767548719 Chr.19: 45350517-45350519 on Build GRCh38
rs766510843 Chr.19: 45350571-45350571 on Build GRCh38
rs3916902 Chr.19: 45350652-45350652 on Build GRCh38
rs750413106 Chr.19: 45350678-45350678 on Build GRCh38
rs199826665 Chr.19: 45350622-45350622 on Build GRCh38
rs545300346 Chr.19: 45350621-45350621 on Build GRCh38
rs762601009 Chr.19: 45350634-45350645 on Build GRCh38
rs755980703 Chr.19: 45350525-45350526 on Build GRCh38
rs201950695 Chr.19: 45350647-45350647 on Build GRCh38
rs773883702 Chr.19: 45350682-45350683 on Build GRCh38
rs375570963 Chr.19: 45350587-45350587 on Build GRCh38
rs373196157 Chr.19: 45350674-45350674 on Build GRCh38
rs568420095 Chr.19: 45350673-45350674 on Build GRCh38
rs565225181 Chr.19: 45350629-45350629 on Build GRCh38
rs182786149 Chr.19: 45350597-45350597 on Build GRCh38

View Full Product Details