Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12294200-12294439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
UniGene
Hs.511899
Gene Chromosome Location
Chr.6: 12256464-12297194 on Build GRCh38
Transcripts
NM_001168319.1 NM_001955.4 XM_011514330.2 XM_011514331.2 XM_011514332.2 XM_017010331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586183 c.462G>A p.K154K Chr.6: 12294333-12294333 on Build GRCh38
3828955 c.446C>A p.S149Y Chr.6: 12294317-12294317 on Build GRCh38
1204924 c.486A>C p.R162S Chr.6: 12294357-12294357 on Build GRCh38
450427 c.456G>T p.G152G Chr.6: 12294327-12294327 on Build GRCh38
109586 c.434G>A p.G145E Chr.6: 12294305-12294305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763046520 Chr.6: 12294372-12294375 on Build GRCh38
rs753790282 Chr.6: 12294359-12294359 on Build GRCh38
rs759763323 Chr.6: 12294278-12294278 on Build GRCh38
rs201765566 Chr.6: 12294257-12294257 on Build GRCh38
rs770051193 Chr.6: 12294363-12294370 on Build GRCh38
rs756265142 Chr.6: 12294358-12294358 on Build GRCh38
rs767323529 Chr.6: 12294349-12294349 on Build GRCh38
rs537170013 Chr.6: 12294353-12294353 on Build GRCh38
rs1019227124 Chr.6: 12294256-12294256 on Build GRCh38
rs765372578 Chr.6: 12294386-12294386 on Build GRCh38
rs748326404 Chr.6: 12294274-12294274 on Build GRCh38
rs951534547 Chr.6: 12294352-12294352 on Build GRCh38
rs759606950 Chr.6: 12294237-12294238 on Build GRCh38
rs150375573 Chr.6: 12294305-12294305 on Build GRCh38
rs778052405 Chr.6: 12294298-12294298 on Build GRCh38
rs766257520 Chr.6: 12294357-12294357 on Build GRCh38
rs983858211 Chr.6: 12294368-12294368 on Build GRCh38
rs777308039 Chr.6: 12294317-12294317 on Build GRCh38
rs149750515 Chr.6: 12294301-12294301 on Build GRCh38
rs149316725 Chr.6: 12294388-12294388 on Build GRCh38
rs901425551 Chr.6: 12294313-12294313 on Build GRCh38
rs774861385 Chr.6: 12294248-12294248 on Build GRCh38
rs1006883209 Chr.6: 12294229-12294229 on Build GRCh38
rs755076473 Chr.6: 12294259-12294259 on Build GRCh38
rs773985828 Chr.6: 12294343-12294343 on Build GRCh38
rs576837257 Chr.6: 12294411-12294411 on Build GRCh38
rs568430735 Chr.6: 12294275-12294275 on Build GRCh38
rs754988566 Chr.6: 12294375-12294375 on Build GRCh38
rs145546137 Chr.6: 12294351-12294351 on Build GRCh38
rs771602064 Chr.6: 12294314-12294314 on Build GRCh38
rs768294608 Chr.6: 12294335-12294335 on Build GRCh38
rs748806941 Chr.6: 12294370-12294372 on Build GRCh38
rs764524594 Chr.6: 12294404-12294404 on Build GRCh38
rs758593055 Chr.6: 12294410-12294410 on Build GRCh38
rs753876378 Chr.6: 12294237-12294237 on Build GRCh38
rs376346685 Chr.6: 12294348-12294348 on Build GRCh38
rs779053288 Chr.6: 12294272-12294272 on Build GRCh38
rs762501115 Chr.6: 12294328-12294328 on Build GRCh38

View Full Product Details