Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19309803-19310076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN2
Entrez Gene ID
22905
Gene Name
epsin 2
Gene Aliases
EHB21
UniGene
Hs.743983
Gene Chromosome Location
Chr.17: 19237377-19336715 on Build GRCh38
Transcripts
NM_001102664.1 NM_014964.4 NM_148921.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6223218 c.839T>C p.L280P Chr.17: 19309957-19309957 on Build GRCh38
976380 c.778C>T p.R260* Chr.17: 19309896-19309896 on Build GRCh38
4766708 c.808C>A p.P270T Chr.17: 19309926-19309926 on Build GRCh38
6655370 c.771C>T p.T257T Chr.17: 19309889-19309889 on Build GRCh38
160810 c.806G>C p.R269P Chr.17: 19309924-19309924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377333169 Chr.17: 19309897-19309897 on Build GRCh38
rs746115625 Chr.17: 19309835-19309835 on Build GRCh38
rs148625960 Chr.17: 19309894-19309894 on Build GRCh38
rs909575750 Chr.17: 19309863-19309863 on Build GRCh38
rs759017700 Chr.17: 19310021-19310021 on Build GRCh38
rs775202472 Chr.17: 19310005-19310005 on Build GRCh38
rs762316060 Chr.17: 19310001-19310001 on Build GRCh38
rs781336503 Chr.17: 19309903-19309903 on Build GRCh38
rs942656645 Chr.17: 19310031-19310031 on Build GRCh38
rs867769180 Chr.17: 19310053-19310053 on Build GRCh38
rs939327997 Chr.17: 19309960-19309960 on Build GRCh38
rs1043982925 Chr.17: 19310023-19310023 on Build GRCh38
rs772330921 Chr.17: 19309956-19309957 on Build GRCh38
rs758310344 Chr.17: 19309892-19309892 on Build GRCh38
rs756361975 Chr.17: 19309877-19309877 on Build GRCh38
rs765930708 Chr.17: 19309873-19309873 on Build GRCh38
rs752519675 Chr.17: 19309891-19309891 on Build GRCh38
rs746757737 Chr.17: 19310019-19310019 on Build GRCh38
rs142086913 Chr.17: 19309910-19309910 on Build GRCh38
rs769071495 Chr.17: 19309940-19309940 on Build GRCh38
rs768479656 Chr.17: 19310032-19310033 on Build GRCh38
rs753368595 Chr.17: 19309876-19309876 on Build GRCh38
rs780186428 Chr.17: 19309908-19309908 on Build GRCh38
rs918337039 Chr.17: 19309925-19309925 on Build GRCh38
rs532861601 Chr.17: 19310013-19310013 on Build GRCh38
rs962635784 Chr.17: 19309833-19309833 on Build GRCh38
rs762828636 Chr.17: 19310045-19310045 on Build GRCh38
rs764894719 Chr.17: 19309888-19309888 on Build GRCh38
rs369980154 Chr.17: 19309907-19309907 on Build GRCh38
rs751475132 Chr.17: 19310048-19310048 on Build GRCh38
rs775952637 Chr.17: 19309846-19309846 on Build GRCh38
rs776230453 Chr.17: 19310019-19310019 on Build GRCh38
rs746132884 Chr.17: 19309860-19309862 on Build GRCh38
rs200526844 Chr.17: 19310046-19310046 on Build GRCh38
rs2296981 Chr.17: 19310022-19310022 on Build GRCh38
rs373826021 Chr.17: 19309858-19309858 on Build GRCh38
rs201108124 Chr.17: 19309861-19309861 on Build GRCh38
rs201275040 Chr.17: 19309991-19309991 on Build GRCh38
rs933790970 Chr.17: 19309943-19309943 on Build GRCh38
rs752464516 Chr.17: 19310024-19310024 on Build GRCh38
rs754692957 Chr.17: 19309881-19309881 on Build GRCh38
rs751606349 Chr.17: 19309896-19309896 on Build GRCh38
rs760169494 Chr.17: 19309867-19309867 on Build GRCh38
rs763399561 Chr.17: 19309856-19309856 on Build GRCh38

View Full Product Details