Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 157136774-157136999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRB
Entrez Gene ID
2743
Gene Name
glycine receptor beta
Gene Aliases
HKPX2
UniGene
Hs.32973
Gene Chromosome Location
Chr.4: 157076125-157172090 on Build GRCh38
Transcripts
NM_000824.4 NM_001166060.1 NM_001166061.1 XM_017008034.1 XM_017008035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6005894 c.548G>T p.C183F Chr.4: 157136824-157136824 on Build GRCh38
1052567 c.608G>T p.S203I Chr.4: 157136884-157136884 on Build GRCh38
1052566 c.587G>C p.R196P Chr.4: 157136863-157136863 on Build GRCh38
7316351 c.564A>G p.T188T Chr.4: 157136840-157136840 on Build GRCh38
6099585 c.601C>A p.L201M Chr.4: 157136877-157136877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780481695 Chr.4: 157136895-157136895 on Build GRCh38
rs554521200 Chr.4: 157136909-157136909 on Build GRCh38
rs770847089 Chr.4: 157136924-157136924 on Build GRCh38
rs770664655 Chr.4: 157136816-157136816 on Build GRCh38
rs936875665 Chr.4: 157136966-157136966 on Build GRCh38
rs398122856 Chr.4: 157136872-157136872 on Build GRCh38
rs1001867844 Chr.4: 157136807-157136807 on Build GRCh38
rs141976916 Chr.4: 157136863-157136863 on Build GRCh38
rs1035041552 Chr.4: 157136836-157136836 on Build GRCh38
rs779156561 Chr.4: 157136890-157136890 on Build GRCh38
rs768659666 Chr.4: 157136908-157136908 on Build GRCh38
rs771827425 Chr.4: 157136935-157136935 on Build GRCh38
rs747212960 Chr.4: 157136904-157136904 on Build GRCh38
rs281864922 Chr.4: 157136891-157136891 on Build GRCh38
rs754330008 Chr.4: 157136877-157136877 on Build GRCh38
rs201716331 Chr.4: 157136936-157136936 on Build GRCh38
rs975522417 Chr.4: 157136862-157136862 on Build GRCh38
rs536787475 Chr.4: 157136883-157136883 on Build GRCh38
rs569240499 Chr.4: 157136837-157136837 on Build GRCh38
rs762918908 Chr.4: 157136810-157136810 on Build GRCh38
rs749207535 Chr.4: 157136911-157136911 on Build GRCh38
rs752345500 Chr.4: 157136843-157136843 on Build GRCh38
rs767093958 Chr.4: 157136842-157136842 on Build GRCh38
rs772673792 Chr.4: 157136926-157136926 on Build GRCh38
rs758829852 Chr.4: 157136893-157136893 on Build GRCh38
rs745734462 Chr.4: 157136929-157136929 on Build GRCh38
rs759202495 Chr.4: 157136824-157136824 on Build GRCh38
rs551200623 Chr.4: 157136822-157136822 on Build GRCh38
rs761029183 Chr.4: 157136825-157136825 on Build GRCh38
rs760224325 Chr.4: 157136856-157136856 on Build GRCh38
rs142603409 Chr.4: 157136967-157136967 on Build GRCh38

View Full Product Details