Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24055482-24055737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2S3
Entrez Gene ID
1968
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Gene Aliases
EIF2, EIF2G, EIF2gamma, eIF-2gA
UniGene
Hs.539684
Gene Chromosome Location
Chr.X: 24054948-24078810 on Build GRCh38
Transcripts
NM_001415.3

SNP Detail

SNP ID Chromosome Location
rs1035645440 Chr.X: 24055688-24055688 on Build GRCh38
rs752318073 Chr.X: 24055654-24055654 on Build GRCh38
rs780705102 Chr.X: 24055608-24055608 on Build GRCh38
rs749156562 Chr.X: 24055657-24055657 on Build GRCh38
rs769932748 Chr.X: 24055669-24055669 on Build GRCh38
rs749986597 Chr.X: 24055591-24055591 on Build GRCh38
rs1025410253 Chr.X: 24055527-24055527 on Build GRCh38
rs1013479065 Chr.X: 24055525-24055525 on Build GRCh38
rs770654105 Chr.X: 24055643-24055643 on Build GRCh38
rs751161180 Chr.X: 24055584-24055584 on Build GRCh38
rs765813361 Chr.X: 24055580-24055580 on Build GRCh38
rs939700278 Chr.X: 24055606-24055606 on Build GRCh38
rs56229020 Chr.X: 24055668-24055668 on Build GRCh38
rs377239918 Chr.X: 24055686-24055686 on Build GRCh38
rs368974008 Chr.X: 24055573-24055573 on Build GRCh38
rs1032427025 Chr.X: 24055546-24055546 on Build GRCh38
rs978587206 Chr.X: 24055543-24055543 on Build GRCh38
rs1002221472 Chr.X: 24055684-24055684 on Build GRCh38
rs777680953 Chr.X: 24055639-24055639 on Build GRCh38
rs113645105 Chr.X: 24055566-24055566 on Build GRCh38
rs974869796 Chr.X: 24055549-24055549 on Build GRCh38
rs754330241 Chr.X: 24055564-24055564 on Build GRCh38
rs749138063 Chr.X: 24055642-24055642 on Build GRCh38
rs779869927 Chr.X: 24055583-24055583 on Build GRCh38
rs36018672 Chr.X: 24055644-24055644 on Build GRCh38
rs201953012 Chr.X: 24055660-24055660 on Build GRCh38
rs776243738 Chr.X: 24055625-24055625 on Build GRCh38
rs1022356975 Chr.X: 24055588-24055588 on Build GRCh38
rs769810966 Chr.X: 24055638-24055638 on Build GRCh38
rs755605721 Chr.X: 24055656-24055656 on Build GRCh38
rs16997658 Chr.X: 24055559-24055559 on Build GRCh38
rs765698724 Chr.X: 24055569-24055570 on Build GRCh38
rs766811075 Chr.X: 24055652-24055652 on Build GRCh38
rs775652363 Chr.X: 24055656-24055656 on Build GRCh38
rs761797523 Chr.X: 24055648-24055648 on Build GRCh38
rs371630608 Chr.X: 24055581-24055581 on Build GRCh38
rs765200746 Chr.X: 24055542-24055542 on Build GRCh38
rs905176810 Chr.X: 24055609-24055609 on Build GRCh38
rs765904960 Chr.X: 24055650-24055650 on Build GRCh38
rs894950891 Chr.X: 24055704-24055704 on Build GRCh38
rs767048198 Chr.X: 24055653-24055653 on Build GRCh38
rs763418298 Chr.X: 24055651-24055651 on Build GRCh38
rs763506053 Chr.X: 24055701-24055701 on Build GRCh38
rs778626122 Chr.X: 24055658-24055658 on Build GRCh38
rs958234922 Chr.X: 24055548-24055548 on Build GRCh38
rs768638930 Chr.X: 24055647-24055647 on Build GRCh38
rs747590626 Chr.X: 24055610-24055610 on Build GRCh38

View Full Product Details