Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31426957-31427230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
UniGene
Hs.115242
Gene Chromosome Location
Chr.22: 31399553-31434186 on Build GRCh38
Transcripts
NM_004147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2938176 c.947C>G p.T316S Chr.22: 31427125-31427125 on Build GRCh38
2938178 c.994G>A p.E332K Chr.22: 31427172-31427172 on Build GRCh38
7222270 c.1004+9C>T p.? Chr.22: 31427191-31427191 on Build GRCh38
7196697 c.974T>A p.I325N Chr.22: 31427152-31427152 on Build GRCh38
4103577 c.947C>T p.T316I Chr.22: 31427125-31427125 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745483561 Chr.22: 31427080-31427080 on Build GRCh38
rs760867674 Chr.22: 31427192-31427192 on Build GRCh38
rs940327256 Chr.22: 31427122-31427122 on Build GRCh38
rs146214852 Chr.22: 31427136-31427136 on Build GRCh38
rs764329393 Chr.22: 31427128-31427128 on Build GRCh38
rs748115183 Chr.22: 31427029-31427029 on Build GRCh38
rs773853780 Chr.22: 31427190-31427190 on Build GRCh38
rs772642640 Chr.22: 31427138-31427138 on Build GRCh38
rs186773573 Chr.22: 31427108-31427108 on Build GRCh38
rs142301532 Chr.22: 31426993-31426993 on Build GRCh38
rs967377904 Chr.22: 31426987-31426987 on Build GRCh38
rs778540888 Chr.22: 31427026-31427026 on Build GRCh38
rs569718179 Chr.22: 31427056-31427056 on Build GRCh38
rs1036240247 Chr.22: 31427125-31427125 on Build GRCh38
rs766692132 Chr.22: 31427011-31427011 on Build GRCh38
rs746447155 Chr.22: 31427048-31427048 on Build GRCh38
rs201137214 Chr.22: 31427121-31427121 on Build GRCh38
rs28668621 Chr.22: 31426990-31426990 on Build GRCh38
rs151287237 Chr.22: 31427003-31427003 on Build GRCh38
rs754129324 Chr.22: 31427196-31427196 on Build GRCh38
rs1040269076 Chr.22: 31427034-31427034 on Build GRCh38
rs776399080 Chr.22: 31427078-31427078 on Build GRCh38
rs369718031 Chr.22: 31427043-31427043 on Build GRCh38
rs375558007 Chr.22: 31427017-31427017 on Build GRCh38
rs746829242 Chr.22: 31427027-31427027 on Build GRCh38
rs762427310 Chr.22: 31427120-31427120 on Build GRCh38
rs758281050 Chr.22: 31427030-31427030 on Build GRCh38
rs143337002 Chr.22: 31427092-31427092 on Build GRCh38
rs753994601 Chr.22: 31427014-31427014 on Build GRCh38
rs975570022 Chr.22: 31427072-31427072 on Build GRCh38

View Full Product Details