Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42077246-42077471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM109B
Entrez Gene ID
150368
Gene Name
family with sequence similarity 109 member B
Gene Aliases
IPIP27B, Ses2
UniGene
Hs.368312
Gene Chromosome Location
Chr.22: 42074244-42079438 on Build GRCh38
Transcripts
NM_001002034.2 XM_005261373.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6162118 c.82G>C p.G28R Chr.22: 42077375-42077375 on Build GRCh38
7319115 c.66C>T p.G22G Chr.22: 42077359-42077359 on Build GRCh38
4653516 c.52G>A p.A18T Chr.22: 42077345-42077345 on Build GRCh38
6332987 c.74G>T p.R25L Chr.22: 42077367-42077367 on Build GRCh38
1034731 c.122G>A p.R41Q Chr.22: 42077415-42077415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765983868 Chr.22: 42077409-42077409 on Build GRCh38
rs973097581 Chr.22: 42077412-42077412 on Build GRCh38
rs771090609 Chr.22: 42077378-42077378 on Build GRCh38
rs368590052 Chr.22: 42077424-42077424 on Build GRCh38
rs143331054 Chr.22: 42077395-42077395 on Build GRCh38
rs139563105 Chr.22: 42077320-42077320 on Build GRCh38
rs975277337 Chr.22: 42077415-42077415 on Build GRCh38
rs1025302969 Chr.22: 42077335-42077335 on Build GRCh38
rs370983239 Chr.22: 42077277-42077277 on Build GRCh38
rs898251781 Chr.22: 42077312-42077312 on Build GRCh38
rs370007312 Chr.22: 42077310-42077310 on Build GRCh38
rs764753406 Chr.22: 42077398-42077398 on Build GRCh38
rs781024582 Chr.22: 42077366-42077366 on Build GRCh38
rs779838091 Chr.22: 42077340-42077340 on Build GRCh38
rs746990767 Chr.22: 42077345-42077345 on Build GRCh38
rs750142577 Chr.22: 42077399-42077399 on Build GRCh38
rs1014080907 Chr.22: 42077373-42077373 on Build GRCh38
rs746760357 Chr.22: 42077272-42077272 on Build GRCh38
rs867099633 Chr.22: 42077324-42077324 on Build GRCh38
rs199870271 Chr.22: 42077422-42077422 on Build GRCh38
rs748433130 Chr.22: 42077344-42077364 on Build GRCh38
rs570916838 Chr.22: 42077278-42077278 on Build GRCh38
rs1034431433 Chr.22: 42077371-42077371 on Build GRCh38
rs772548726 Chr.22: 42077382-42077382 on Build GRCh38
rs199537712 Chr.22: 42077305-42077305 on Build GRCh38
rs576764942 Chr.22: 42077325-42077325 on Build GRCh38
rs200670447 Chr.22: 42077306-42077306 on Build GRCh38
rs775937104 Chr.22: 42077393-42077393 on Build GRCh38
rs1019503876 Chr.22: 42077394-42077394 on Build GRCh38
rs367573147 Chr.22: 42077328-42077328 on Build GRCh38
rs879219027 Chr.22: 42077397-42077397 on Build GRCh38
rs951282554 Chr.22: 42077347-42077347 on Build GRCh38
rs769841844 Chr.22: 42077367-42077367 on Build GRCh38
rs962247484 Chr.22: 42077400-42077400 on Build GRCh38
rs772414209 Chr.22: 42077372-42077373 on Build GRCh38
rs753535017 Chr.22: 42077313-42077313 on Build GRCh38
rs74337819 Chr.22: 42077284-42077284 on Build GRCh38
rs371373425 Chr.22: 42077410-42077410 on Build GRCh38
rs755960326 Chr.22: 42077437-42077437 on Build GRCh38
rs200559139 Chr.22: 42077311-42077311 on Build GRCh38
rs773515466 Chr.22: 42077372-42077372 on Build GRCh38
rs774965754 Chr.22: 42077379-42077379 on Build GRCh38
rs986708202 Chr.22: 42077377-42077377 on Build GRCh38
rs754766889 Chr.22: 42077346-42077346 on Build GRCh38
rs985881556 Chr.22: 42077444-42077444 on Build GRCh38
rs757951633 Chr.22: 42077334-42077334 on Build GRCh38
rs752449621 Chr.22: 42077434-42077434 on Build GRCh38
rs375692639 Chr.22: 42077295-42077295 on Build GRCh38
rs764954100 Chr.22: 42077327-42077327 on Build GRCh38
rs749276957 Chr.22: 42077376-42077376 on Build GRCh38
rs747018766 Chr.22: 42077447-42077448 on Build GRCh38
rs779119019 Chr.22: 42077308-42077310 on Build GRCh38
rs762343043 Chr.22: 42077406-42077406 on Build GRCh38
rs200775641 Chr.22: 42077373-42077373 on Build GRCh38
rs773109898 Chr.22: 42077299-42077299 on Build GRCh38

View Full Product Details