Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94949033-94949282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C9
Entrez Gene ID
1559
Gene Name
cytochrome P450 family 2 subfamily C member 9
Gene Aliases
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
UniGene
Hs.282624
Gene Chromosome Location
Chr.10: 94905867-94989395 on Build GRCh38
Transcripts
NM_000771.3 XM_017015758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639217 c.697T>G p.L233V Chr.10: 94949162-94949162 on Build GRCh38
5927578 c.778C>T p.P260S Chr.10: 94949243-94949243 on Build GRCh38
4455424 c.767A>C p.D256A Chr.10: 94949232-94949232 on Build GRCh38
6639219 c.653A>T p.N218I Chr.10: 94949118-94949118 on Build GRCh38
5932582 c.776A>G p.N259S Chr.10: 94949241-94949241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761086673 Chr.10: 94949153-94949153 on Build GRCh38
rs781030703 Chr.10: 94949196-94949196 on Build GRCh38
rs769077255 Chr.10: 94949247-94949247 on Build GRCh38
rs765176937 Chr.10: 94949173-94949173 on Build GRCh38
rs369745682 Chr.10: 94949184-94949184 on Build GRCh38
rs755804202 Chr.10: 94949218-94949218 on Build GRCh38
rs747588944 Chr.10: 94949242-94949242 on Build GRCh38
rs560483134 Chr.10: 94949078-94949078 on Build GRCh38
rs779117843 Chr.10: 94949073-94949073 on Build GRCh38
rs759708513 Chr.10: 94949107-94949107 on Build GRCh38
rs780801862 Chr.10: 94949118-94949118 on Build GRCh38
rs549167718 Chr.10: 94949172-94949172 on Build GRCh38
rs761895497 Chr.10: 94949256-94949256 on Build GRCh38
rs751549130 Chr.10: 94949178-94949178 on Build GRCh38
rs2256871 Chr.10: 94949217-94949217 on Build GRCh38
rs769180786 Chr.10: 94949140-94949140 on Build GRCh38
rs780013210 Chr.10: 94949176-94949176 on Build GRCh38
rs758372728 Chr.10: 94949097-94949097 on Build GRCh38
rs779666293 Chr.10: 94949103-94949103 on Build GRCh38
rs764614062 Chr.10: 94949155-94949155 on Build GRCh38
rs754111693 Chr.10: 94949169-94949169 on Build GRCh38
rs945414728 Chr.10: 94949213-94949213 on Build GRCh38
rs775108214 Chr.10: 94949202-94949202 on Build GRCh38
rs776115064 Chr.10: 94949241-94949241 on Build GRCh38
rs777152877 Chr.10: 94949252-94949252 on Build GRCh38
rs1042378453 Chr.10: 94949216-94949216 on Build GRCh38
rs747855005 Chr.10: 94949138-94949138 on Build GRCh38
rs1003768765 Chr.10: 94949231-94949231 on Build GRCh38
rs895080856 Chr.10: 94949148-94949148 on Build GRCh38
rs769753119 Chr.10: 94949069-94949070 on Build GRCh38
rs775955387 Chr.10: 94949150-94949150 on Build GRCh38
rs1054686967 Chr.10: 94949195-94949195 on Build GRCh38
rs915366633 Chr.10: 94949201-94949201 on Build GRCh38
rs748856817 Chr.10: 94949229-94949229 on Build GRCh38
rs1055330320 Chr.10: 94949244-94949244 on Build GRCh38
rs902312425 Chr.10: 94949243-94949243 on Build GRCh38
rs377065367 Chr.10: 94949096-94949096 on Build GRCh38
rs750539902 Chr.10: 94949174-94949174 on Build GRCh38
rs867521587 Chr.10: 94949131-94949131 on Build GRCh38
rs770384822 Chr.10: 94949238-94949238 on Build GRCh38
rs149147712 Chr.10: 94949062-94949062 on Build GRCh38
rs777095173 Chr.10: 94949222-94949222 on Build GRCh38
rs758229045 Chr.10: 94949175-94949175 on Build GRCh38
rs772651628 Chr.10: 94949146-94949146 on Build GRCh38
rs772782449 Chr.10: 94949145-94949145 on Build GRCh38
rs922228767 Chr.10: 94949090-94949090 on Build GRCh38
rs199841629 Chr.10: 94949186-94949186 on Build GRCh38

View Full Product Details