Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150511542-150511775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECM1
Entrez Gene ID
1893
Gene Name
extracellular matrix protein 1
Gene Aliases
URBWD
UniGene
Hs.81071
Gene Chromosome Location
Chr.1: 150508011-150513789 on Build GRCh38
Transcripts
NM_001202858.1 NM_004425.3 NM_022664.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622100 c.937A>G p.S313G Chr.1: 150511604-150511604 on Build GRCh38
3473433 c.875C>T p.S292F Chr.1: 150511623-150511623 on Build GRCh38
3473434 c.978C>T p.F326F Chr.1: 150511645-150511645 on Build GRCh38
5530322 c.912C>T p.P304P Chr.1: 150511660-150511660 on Build GRCh38
4458112 c.990C>T p.P330P Chr.1: 150511738-150511738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756418996 Chr.1: 150511656-150511656 on Build GRCh38
rs1050000707 Chr.1: 150511660-150511660 on Build GRCh38
rs757995294 Chr.1: 150511672-150511672 on Build GRCh38
rs748043488 Chr.1: 150511697-150511697 on Build GRCh38
rs781194369 Chr.1: 150511737-150511737 on Build GRCh38
rs766097780 Chr.1: 150511728-150511728 on Build GRCh38
rs750025579 Chr.1: 150511671-150511671 on Build GRCh38
rs754444452 Chr.1: 150511735-150511735 on Build GRCh38
rs148594423 Chr.1: 150511715-150511715 on Build GRCh38
rs774865953 Chr.1: 150511640-150511640 on Build GRCh38
rs80339946 Chr.1: 150511568-150511568 on Build GRCh38
rs760051136 Chr.1: 150511641-150511641 on Build GRCh38
rs750123295 Chr.1: 150511723-150511723 on Build GRCh38
rs757974844 Chr.1: 150511562-150511562 on Build GRCh38
rs377089003 Chr.1: 150511622-150511622 on Build GRCh38
rs377048885 Chr.1: 150511725-150511725 on Build GRCh38
rs754982050 Chr.1: 150511695-150511695 on Build GRCh38
rs869025563 Chr.1: 150511574-150511574 on Build GRCh38
rs747875899 Chr.1: 150511611-150511611 on Build GRCh38
rs775854728 Chr.1: 150511718-150511718 on Build GRCh38
rs587707384 Chr.1: 150511748-150511748 on Build GRCh38
rs199759513 Chr.1: 150511650-150511650 on Build GRCh38
rs587775247 Chr.1: 150511749-150511749 on Build GRCh38
rs771034233 Chr.1: 150511703-150511703 on Build GRCh38
rs751140798 Chr.1: 150511734-150511734 on Build GRCh38
rs768125316 Chr.1: 150511716-150511716 on Build GRCh38
rs201302483 Chr.1: 150511738-150511738 on Build GRCh38
rs139981276 Chr.1: 150511704-150511704 on Build GRCh38
rs766392166 Chr.1: 150511627-150511627 on Build GRCh38
rs761101717 Chr.1: 150511720-150511720 on Build GRCh38
rs764491442 Chr.1: 150511721-150511721 on Build GRCh38
rs768356932 Chr.1: 150511592-150511592 on Build GRCh38
rs765003612 Chr.1: 150511657-150511657 on Build GRCh38
rs779661460 Chr.1: 150511689-150511689 on Build GRCh38
rs143016855 Chr.1: 150511593-150511593 on Build GRCh38
rs780144230 Chr.1: 150511570-150511570 on Build GRCh38
rs910060517 Chr.1: 150511613-150511613 on Build GRCh38
rs368554930 Chr.1: 150511617-150511617 on Build GRCh38
rs951560097 Chr.1: 150511564-150511564 on Build GRCh38
rs78332481 Chr.1: 150511641-150511641 on Build GRCh38
rs780980541 Chr.1: 150511696-150511696 on Build GRCh38
rs746885747 Chr.1: 150511581-150511581 on Build GRCh38
rs771623652 Chr.1: 150511743-150511743 on Build GRCh38
rs369747136 Chr.1: 150511626-150511626 on Build GRCh38
rs769409491 Chr.1: 150511698-150511698 on Build GRCh38
rs753152087 Chr.1: 150511651-150511651 on Build GRCh38
rs746971908 Chr.1: 150511691-150511691 on Build GRCh38
rs767882878 Chr.1: 150511645-150511645 on Build GRCh38

View Full Product Details