Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43774725-43774968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP3
Entrez Gene ID
1845
Gene Name
dual specificity phosphatase 3
Gene Aliases
VHR
UniGene
Hs.181046
Gene Chromosome Location
Chr.17: 43766121-43779000 on Build GRCh38
Transcripts
NM_004090.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5699597 c.145C>T p.P49S Chr.17: 43774919-43774919 on Build GRCh38
7427353 c.261C>T p.G87G Chr.17: 43774803-43774803 on Build GRCh38
5981232 c.149A>G p.K50R Chr.17: 43774915-43774915 on Build GRCh38
7294414 c.207G>A p.M69I Chr.17: 43774857-43774857 on Build GRCh38
5386437 c.290A>G p.N97S Chr.17: 43774774-43774774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998172940 Chr.17: 43774888-43774888 on Build GRCh38
rs972411872 Chr.17: 43774772-43774772 on Build GRCh38
rs1028846665 Chr.17: 43774892-43774892 on Build GRCh38
rs761819392 Chr.17: 43774824-43774824 on Build GRCh38
rs377024727 Chr.17: 43774917-43774917 on Build GRCh38
rs149560689 Chr.17: 43774812-43774812 on Build GRCh38
rs146116274 Chr.17: 43774791-43774791 on Build GRCh38
rs890748021 Chr.17: 43774813-43774813 on Build GRCh38
rs779417821 Chr.17: 43774753-43774753 on Build GRCh38
rs756294930 Chr.17: 43774889-43774889 on Build GRCh38
rs763059564 Chr.17: 43774818-43774818 on Build GRCh38
rs781233840 Chr.17: 43774880-43774880 on Build GRCh38
rs144239960 Chr.17: 43774843-43774843 on Build GRCh38
rs753173506 Chr.17: 43774762-43774762 on Build GRCh38
rs1004757835 Chr.17: 43774890-43774890 on Build GRCh38
rs774103710 Chr.17: 43774821-43774821 on Build GRCh38
rs142204195 Chr.17: 43774787-43774787 on Build GRCh38
rs758652091 Chr.17: 43774856-43774856 on Build GRCh38
rs747583349 Chr.17: 43774785-43774785 on Build GRCh38
rs369878372 Chr.17: 43774845-43774845 on Build GRCh38
rs781254028 Chr.17: 43774788-43774788 on Build GRCh38
rs768275135 Chr.17: 43774915-43774915 on Build GRCh38
rs774894685 Chr.17: 43774854-43774854 on Build GRCh38
rs780207572 Chr.17: 43774899-43774899 on Build GRCh38
rs751943754 Chr.17: 43774878-43774878 on Build GRCh38
rs868036321 Chr.17: 43774804-43774804 on Build GRCh38
rs757849378 Chr.17: 43774879-43774879 on Build GRCh38
rs749543145 Chr.17: 43774902-43774902 on Build GRCh38
rs372866838 Chr.17: 43774875-43774875 on Build GRCh38
rs562230366 Chr.17: 43774820-43774820 on Build GRCh38
rs758918021 Chr.17: 43774766-43774766 on Build GRCh38
rs778185056 Chr.17: 43774776-43774776 on Build GRCh38
rs138980602 Chr.17: 43774792-43774792 on Build GRCh38
rs148312361 Chr.17: 43774881-43774881 on Build GRCh38
rs374962096 Chr.17: 43774746-43774746 on Build GRCh38
rs766186708 Chr.17: 43774847-43774847 on Build GRCh38

View Full Product Details