Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232556363-232556630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E2
Entrez Gene ID
9470
Gene Name
eukaryotic translation initiation factor 4E family member 2
Gene Aliases
4E-LP, 4EHP, EIF4EL3, IF4e
UniGene
Hs.292026
Gene Chromosome Location
Chr.2: 232550587-232583645 on Build GRCh38
Transcripts
NM_001276336.1 NM_001276337.1 NM_001282958.1 NM_004846.3 XM_005246975.3 XM_011512206.2 XM_017005377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619790 c.34G>A p.D12N Chr.2: 232556429-232556429 on Build GRCh38
1017845 c.121A>G p.S41G Chr.2: 232556516-232556516 on Build GRCh38
5463102 c.134A>C p.K45T Chr.2: 232556529-232556529 on Build GRCh38
4092413 c.99G>A p.T33T Chr.2: 232556494-232556494 on Build GRCh38
6157649 c.28G>A p.D10N Chr.2: 232556423-232556423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757321950 Chr.2: 232556464-232556464 on Build GRCh38
rs763258335 Chr.2: 232556417-232556417 on Build GRCh38
rs557337169 Chr.2: 232556455-232556455 on Build GRCh38
rs201272768 Chr.2: 232556575-232556575 on Build GRCh38
rs3830645 Chr.2: 232556575-232556575 on Build GRCh38
rs370171571 Chr.2: 232556498-232556498 on Build GRCh38
rs113235222 Chr.2: 232556580-232556580 on Build GRCh38
rs397740637 Chr.2: 232556578-232556578 on Build GRCh38
rs747719438 Chr.2: 232556504-232556504 on Build GRCh38
rs780492980 Chr.2: 232556494-232556494 on Build GRCh38
rs969802613 Chr.2: 232556525-232556525 on Build GRCh38
rs993859399 Chr.2: 232556548-232556548 on Build GRCh38
rs777132374 Chr.2: 232556578-232556579 on Build GRCh38
rs770724027 Chr.2: 232556555-232556555 on Build GRCh38
rs537332303 Chr.2: 232556392-232556392 on Build GRCh38
rs776499574 Chr.2: 232556577-232556577 on Build GRCh38
rs766167787 Chr.2: 232556394-232556394 on Build GRCh38
rs1026240782 Chr.2: 232556567-232556567 on Build GRCh38
rs764241412 Chr.2: 232556447-232556447 on Build GRCh38
rs768067503 Chr.2: 232556485-232556485 on Build GRCh38
rs773064609 Chr.2: 232556545-232556545 on Build GRCh38
rs750915667 Chr.2: 232556492-232556492 on Build GRCh38
rs774540905 Chr.2: 232556582-232556582 on Build GRCh38
rs573880937 Chr.2: 232556590-232556590 on Build GRCh38
rs374932592 Chr.2: 232556552-232556552 on Build GRCh38
rs778707309 Chr.2: 232556500-232556500 on Build GRCh38
rs552554328 Chr.2: 232556554-232556554 on Build GRCh38
rs374300851 Chr.2: 232556475-232556475 on Build GRCh38
rs377735542 Chr.2: 232556493-232556493 on Build GRCh38
rs750359341 Chr.2: 232556542-232556546 on Build GRCh38
rs962653205 Chr.2: 232556596-232556596 on Build GRCh38
rs771687914 Chr.2: 232556518-232556518 on Build GRCh38
rs764365992 Chr.2: 232556581-232556581 on Build GRCh38
rs149079029 Chr.2: 232556499-232556499 on Build GRCh38
rs200634717 Chr.2: 232556487-232556487 on Build GRCh38
rs781649020 Chr.2: 232556577-232556578 on Build GRCh38
rs776249676 Chr.2: 232556397-232556397 on Build GRCh38

View Full Product Details