Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 162142115-162142370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG2
Entrez Gene ID
2566
Gene Name
gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases
CAE2, ECA2, GEFSP3
UniGene
Hs.7195
Gene Chromosome Location
Chr.5: 162067642-162155539 on Build GRCh38
Transcripts
NM_000816.3 NM_198903.2 NM_198904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5402957 c.904C>T p.P302S Chr.5: 162142298-162142298 on Build GRCh38
339852 c.917C>G p.S306C Chr.5: 162142311-162142311 on Build GRCh38
6478578 c.876G>C p.V292V Chr.5: 162142270-162142270 on Build GRCh38
207493 c.781G>T p.V261F Chr.5: 162142175-162142175 on Build GRCh38
1310911 c.791T>C p.V264A Chr.5: 162142185-162142185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867159750 Chr.5: 162142160-162142160 on Build GRCh38
rs754514348 Chr.5: 162142183-162142183 on Build GRCh38
rs751069405 Chr.5: 162142157-162142157 on Build GRCh38
rs749127891 Chr.5: 162142237-162142237 on Build GRCh38
rs868390429 Chr.5: 162142161-162142161 on Build GRCh38
rs796052507 Chr.5: 162142163-162142163 on Build GRCh38
rs115126975 Chr.5: 162142192-162142192 on Build GRCh38
rs768565280 Chr.5: 162142256-162142256 on Build GRCh38
rs796052508 Chr.5: 162142238-162142238 on Build GRCh38
rs766844183 Chr.5: 162142155-162142155 on Build GRCh38
rs780766516 Chr.5: 162142188-162142188 on Build GRCh38
rs778780107 Chr.5: 162142255-162142255 on Build GRCh38
rs375294947 Chr.5: 162142322-162142322 on Build GRCh38
rs372757018 Chr.5: 162142258-162142258 on Build GRCh38
rs375947335 Chr.5: 162142329-162142329 on Build GRCh38
rs766906928 Chr.5: 162142334-162142334 on Build GRCh38
rs369989217 Chr.5: 162142147-162142147 on Build GRCh38
rs543339576 Chr.5: 162142293-162142293 on Build GRCh38
rs1051704613 Chr.5: 162142339-162142339 on Build GRCh38
rs796052520 Chr.5: 162142202-162142202 on Build GRCh38
rs558779533 Chr.5: 162142234-162142234 on Build GRCh38
rs200449368 Chr.5: 162142204-162142204 on Build GRCh38
rs776876970 Chr.5: 162142285-162142285 on Build GRCh38
rs770181296 Chr.5: 162142319-162142319 on Build GRCh38
rs765653579 Chr.5: 162142146-162142146 on Build GRCh38
rs763486287 Chr.5: 162142148-162142148 on Build GRCh38
rs770884037 Chr.5: 162142249-162142249 on Build GRCh38
rs11575991 Chr.5: 162142336-162142336 on Build GRCh38
rs796052509 Chr.5: 162142313-162142313 on Build GRCh38

View Full Product Details