Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12677065-12677304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHPS
Entrez Gene ID
1725
Gene Name
deoxyhypusine synthase
Gene Aliases
DHS, DS, MIG13
UniGene
Hs.79064
Gene Chromosome Location
Chr.19: 12672164-12681887 on Build GRCh38
Transcripts
NM_001206974.1 NM_001930.3 NM_013406.2 NR_038192.1 XM_011527770.1 XM_011527771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122771 c.847G>T p.G283W Chr.19: 12677149-12677149 on Build GRCh38
991400 c.824G>A p.C275Y Chr.19: 12677172-12677172 on Build GRCh38
4854829 c.800A>G p.N267S Chr.19: 12677196-12677196 on Build GRCh38
6990537 c.888G>C p.M296I Chr.19: 12677108-12677108 on Build GRCh38
7364275 c.834G>T p.M278I Chr.19: 12677162-12677162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368781559 Chr.19: 12677131-12677131 on Build GRCh38
rs370385403 Chr.19: 12677272-12677272 on Build GRCh38
rs376157066 Chr.19: 12677284-12677284 on Build GRCh38
rs770007022 Chr.19: 12677085-12677085 on Build GRCh38
rs189109985 Chr.19: 12677283-12677283 on Build GRCh38
rs575787787 Chr.19: 12677189-12677189 on Build GRCh38
rs758418180 Chr.19: 12677190-12677190 on Build GRCh38
rs764915726 Chr.19: 12677271-12677271 on Build GRCh38
rs978898685 Chr.19: 12677158-12677158 on Build GRCh38
rs767613292 Chr.19: 12677162-12677162 on Build GRCh38
rs749528954 Chr.19: 12677224-12677224 on Build GRCh38
rs756853042 Chr.19: 12677104-12677104 on Build GRCh38
rs755843603 Chr.19: 12677100-12677100 on Build GRCh38
rs763848755 Chr.19: 12677101-12677101 on Build GRCh38
rs745985350 Chr.19: 12677228-12677228 on Build GRCh38
rs765716761 Chr.19: 12677266-12677268 on Build GRCh38
rs974776882 Chr.19: 12677252-12677252 on Build GRCh38
rs568560231 Chr.19: 12677230-12677230 on Build GRCh38
rs181350133 Chr.19: 12677138-12677138 on Build GRCh38
rs747522720 Chr.19: 12677119-12677119 on Build GRCh38
rs774487656 Chr.19: 12677090-12677090 on Build GRCh38
rs760044789 Chr.19: 12677183-12677183 on Build GRCh38
rs912160587 Chr.19: 12677214-12677214 on Build GRCh38
rs781692345 Chr.19: 12677193-12677193 on Build GRCh38
rs371375475 Chr.19: 12677262-12677262 on Build GRCh38
rs934680485 Chr.19: 12677178-12677178 on Build GRCh38
rs753137312 Chr.19: 12677216-12677216 on Build GRCh38
rs565337083 Chr.19: 12677223-12677223 on Build GRCh38
rs377182954 Chr.19: 12677268-12677268 on Build GRCh38
rs140764692 Chr.19: 12677147-12677147 on Build GRCh38
rs759569057 Chr.19: 12677151-12677151 on Build GRCh38
rs770020319 Chr.19: 12677143-12677143 on Build GRCh38
rs749455821 Chr.19: 12677146-12677146 on Build GRCh38
rs373000198 Chr.19: 12677227-12677227 on Build GRCh38
rs748558868 Chr.19: 12677139-12677139 on Build GRCh38
rs112206538 Chr.19: 12677094-12677094 on Build GRCh38
rs761309007 Chr.19: 12677187-12677187 on Build GRCh38
rs149307460 Chr.19: 12677115-12677115 on Build GRCh38
rs201819675 Chr.19: 12677087-12677087 on Build GRCh38
rs774505760 Chr.19: 12677150-12677150 on Build GRCh38
rs925940285 Chr.19: 12677175-12677175 on Build GRCh38
rs1028652737 Chr.19: 12677254-12677254 on Build GRCh38
rs550355399 Chr.19: 12677183-12677183 on Build GRCh38
rs775628435 Chr.19: 12677235-12677235 on Build GRCh38
rs763786266 Chr.19: 12677177-12677177 on Build GRCh38
rs754130948 Chr.19: 12677149-12677150 on Build GRCh38
rs756557667 Chr.19: 12677222-12677222 on Build GRCh38
rs776254007 Chr.19: 12677267-12677267 on Build GRCh38
rs75392871 Chr.19: 12677225-12677225 on Build GRCh38
rs960496806 Chr.19: 12677118-12677118 on Build GRCh38
rs757878296 Chr.19: 12677191-12677191 on Build GRCh38
rs995883582 Chr.19: 12677123-12677123 on Build GRCh38
rs374394052 Chr.19: 12677093-12677093 on Build GRCh38
rs764762126 Chr.19: 12677188-12677188 on Build GRCh38
rs144283239 Chr.19: 12677144-12677144 on Build GRCh38
rs1044659893 Chr.19: 12677127-12677127 on Build GRCh38
rs554629330 Chr.19: 12677088-12677088 on Build GRCh38
rs775538805 Chr.19: 12677167-12677167 on Build GRCh38
rs760586806 Chr.19: 12677168-12677168 on Build GRCh38
rs752493059 Chr.19: 12677095-12677095 on Build GRCh38
rs898516088 Chr.19: 12677103-12677103 on Build GRCh38

View Full Product Details