Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1215988-1216237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP1
Entrez Gene ID
1487
Gene Name
C-terminal binding protein 1
Gene Aliases
BARS
UniGene
Hs.208597
Gene Chromosome Location
Chr.4: 1211440-1250329 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

Gene Symbol
CTBP1-AS
Entrez Gene ID
285463
Gene Name
CTBP1 antisense RNA
Gene Aliases
PCAT10
UniGene
Hs.707614
Gene Chromosome Location
Chr.4: 1210120-1218591 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887245 c.685C>A p.H229N Chr.4: 1216068-1216068 on Build GRCh38
3409031 c.711C>T p.C237C Chr.4: 1216042-1216042 on Build GRCh38
5037231 c.714C>A p.G238G Chr.4: 1216039-1216039 on Build GRCh38
353252 c.570G>T p.A190A Chr.4: 1216183-1216183 on Build GRCh38
1426543 c.637C>T p.R213W Chr.4: 1216116-1216116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778142796 Chr.4: 1216144-1216144 on Build GRCh38
rs369657048 Chr.4: 1216033-1216033 on Build GRCh38
rs757882322 Chr.4: 1216123-1216123 on Build GRCh38
rs747578892 Chr.4: 1216156-1216156 on Build GRCh38
rs374344295 Chr.4: 1216042-1216042 on Build GRCh38
rs147464431 Chr.4: 1216009-1216009 on Build GRCh38
rs756451089 Chr.4: 1216067-1216067 on Build GRCh38
rs746145361 Chr.4: 1216162-1216162 on Build GRCh38
rs772801359 Chr.4: 1216189-1216189 on Build GRCh38
rs774773418 Chr.4: 1216091-1216091 on Build GRCh38
rs142670248 Chr.4: 1216164-1216164 on Build GRCh38
rs765846448 Chr.4: 1216203-1216203 on Build GRCh38
rs745938565 Chr.4: 1216097-1216097 on Build GRCh38
rs370377200 Chr.4: 1216165-1216165 on Build GRCh38
rs375283311 Chr.4: 1216214-1216214 on Build GRCh38
rs779250856 Chr.4: 1216157-1216157 on Build GRCh38
rs753205546 Chr.4: 1216131-1216131 on Build GRCh38
rs764862588 Chr.4: 1216116-1216116 on Build GRCh38
rs371355192 Chr.4: 1216112-1216112 on Build GRCh38
rs746904547 Chr.4: 1216169-1216169 on Build GRCh38
rs768461350 Chr.4: 1216171-1216171 on Build GRCh38
rs776693173 Chr.4: 1216179-1216179 on Build GRCh38
rs750112627 Chr.4: 1216122-1216122 on Build GRCh38
rs757966374 Chr.4: 1216060-1216060 on Build GRCh38
rs932604953 Chr.4: 1216202-1216202 on Build GRCh38
rs374755819 Chr.4: 1216208-1216208 on Build GRCh38
rs139911392 Chr.4: 1216039-1216039 on Build GRCh38
rs376681305 Chr.4: 1216201-1216201 on Build GRCh38
rs778027768 Chr.4: 1216077-1216077 on Build GRCh38
rs761651219 Chr.4: 1216115-1216115 on Build GRCh38
rs781701519 Chr.4: 1216063-1216063 on Build GRCh38
rs1006094126 Chr.4: 1216062-1216062 on Build GRCh38
rs775480221 Chr.4: 1216104-1216104 on Build GRCh38
rs749461434 Chr.4: 1216083-1216083 on Build GRCh38
rs763847829 Chr.4: 1216011-1216011 on Build GRCh38
rs759145559 Chr.4: 1216209-1216209 on Build GRCh38
rs750734000 Chr.4: 1216041-1216041 on Build GRCh38
rs765065949 Chr.4: 1216185-1216185 on Build GRCh38
rs748651372 Chr.4: 1216066-1216066 on Build GRCh38
rs913105300 Chr.4: 1216184-1216184 on Build GRCh38
rs377714722 Chr.4: 1216111-1216111 on Build GRCh38
rs952944457 Chr.4: 1216056-1216056 on Build GRCh38
rs370649780 Chr.4: 1216170-1216170 on Build GRCh38
rs756584802 Chr.4: 1216138-1216138 on Build GRCh38
rs186090455 Chr.4: 1216183-1216183 on Build GRCh38
rs372599555 Chr.4: 1216150-1216150 on Build GRCh38
rs529740572 Chr.4: 1216095-1216095 on Build GRCh38
rs780706169 Chr.4: 1216084-1216084 on Build GRCh38
rs772323252 Chr.4: 1216098-1216098 on Build GRCh38
rs567980949 Chr.4: 1216130-1216130 on Build GRCh38
rs754436909 Chr.4: 1216213-1216213 on Build GRCh38
rs571881989 Chr.4: 1216211-1216211 on Build GRCh38

View Full Product Details