Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148398900-148399171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO38
Entrez Gene ID
81545
Gene Name
F-box protein 38
Gene Aliases
Fbx38, HMN2D, MOKA, SP329
UniGene
Hs.483772
Gene Chromosome Location
Chr.5: 148383935-148442836 on Build GRCh38
Transcripts
NM_001271723.1 NM_030793.4 XM_005268513.1 XM_006714797.1 XM_011537684.2 XM_017009899.1 XM_017009900.1 XM_017009901.1 XM_017009902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336865 c.148A>T p.I50F Chr.5: 148399018-148399018 on Build GRCh38
6586197 c.246G>A p.W82* Chr.5: 148399116-148399116 on Build GRCh38
3211032 c.189A>G p.A63A Chr.5: 148399059-148399059 on Build GRCh38
1642656 c.210T>C p.V70V Chr.5: 148399080-148399080 on Build GRCh38
4141546 c.195C>T p.T65T Chr.5: 148399065-148399065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760555100 Chr.5: 148398965-148398965 on Build GRCh38
rs760456022 Chr.5: 148399043-148399043 on Build GRCh38
rs770050273 Chr.5: 148399001-148399001 on Build GRCh38
rs769047668 Chr.5: 148398979-148398979 on Build GRCh38
rs781388776 Chr.5: 148399127-148399127 on Build GRCh38
rs74863106 Chr.5: 148399068-148399068 on Build GRCh38
rs753296785 Chr.5: 148398973-148398973 on Build GRCh38
rs926850592 Chr.5: 148398931-148398931 on Build GRCh38
rs184238855 Chr.5: 148399136-148399136 on Build GRCh38
rs755427916 Chr.5: 148399110-148399110 on Build GRCh38
rs114174259 Chr.5: 148398933-148398933 on Build GRCh38
rs1056528608 Chr.5: 148398935-148398935 on Build GRCh38
rs372293727 Chr.5: 148398978-148398978 on Build GRCh38
rs758058399 Chr.5: 148399100-148399100 on Build GRCh38
rs761704413 Chr.5: 148399076-148399076 on Build GRCh38
rs775165552 Chr.5: 148399042-148399042 on Build GRCh38
rs748653699 Chr.5: 148398989-148398989 on Build GRCh38
rs192558532 Chr.5: 148398942-148398942 on Build GRCh38
rs764038838 Chr.5: 148398972-148398972 on Build GRCh38
rs779999660 Chr.5: 148399139-148399140 on Build GRCh38
rs951573951 Chr.5: 148398943-148398943 on Build GRCh38
rs771139315 Chr.5: 148399041-148399041 on Build GRCh38
rs530292493 Chr.5: 148398925-148398926 on Build GRCh38
rs756813657 Chr.5: 148398974-148398974 on Build GRCh38
rs369795857 Chr.5: 148398977-148398977 on Build GRCh38
rs749885240 Chr.5: 148399090-148399090 on Build GRCh38
rs752500020 Chr.5: 148398962-148398962 on Build GRCh38
rs781756798 Chr.5: 148398984-148398984 on Build GRCh38
rs751167811 Chr.5: 148399109-148399109 on Build GRCh38
rs996604793 Chr.5: 148399103-148399103 on Build GRCh38
rs370723729 Chr.5: 148398992-148398992 on Build GRCh38
rs375508631 Chr.5: 148399022-148399022 on Build GRCh38
rs778024487 Chr.5: 148399141-148399141 on Build GRCh38
rs764755944 Chr.5: 148399078-148399078 on Build GRCh38
rs748706886 Chr.5: 148399131-148399131 on Build GRCh38
rs763779865 Chr.5: 148399062-148399062 on Build GRCh38
rs78091953 Chr.5: 148398934-148398934 on Build GRCh38
rs765989865 Chr.5: 148399106-148399106 on Build GRCh38
rs756650989 Chr.5: 148399140-148399140 on Build GRCh38
rs773397825 Chr.5: 148399005-148399005 on Build GRCh38
rs376302135 Chr.5: 148398951-148398951 on Build GRCh38

View Full Product Details