Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7246966-7247193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDNEP1
Entrez Gene ID
23399
Gene Name
CTD nuclear envelope phosphatase 1
Gene Aliases
DULLARD, HSA011916, NET56
UniGene
Hs.513913
Gene Chromosome Location
Chr.17: 7243587-7251940 on Build GRCh38
Transcripts
NM_001143775.1 NM_015343.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630378 c.170-5T>C p.? Chr.17: 7247187-7247187 on Build GRCh38
3718512 c.180G>A p.K60K Chr.17: 7247172-7247172 on Build GRCh38
1750307 c.177G>A p.V59V Chr.17: 7247175-7247175 on Build GRCh38
7164044 c.238G>T p.V80F Chr.17: 7247114-7247114 on Build GRCh38
7118562 c.266C>T p.T89M Chr.17: 7247086-7247086 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906033363 Chr.17: 7247042-7247042 on Build GRCh38
rs748905977 Chr.17: 7247067-7247067 on Build GRCh38
rs557493144 Chr.17: 7246998-7246998 on Build GRCh38
rs759399442 Chr.17: 7247086-7247086 on Build GRCh38
rs765316801 Chr.17: 7247105-7247105 on Build GRCh38
rs750607226 Chr.17: 7247018-7247018 on Build GRCh38
rs777180954 Chr.17: 7247060-7247060 on Build GRCh38
rs200165512 Chr.17: 7247130-7247130 on Build GRCh38
rs755315856 Chr.17: 7247157-7247157 on Build GRCh38
rs758576644 Chr.17: 7247027-7247027 on Build GRCh38
rs567687182 Chr.17: 7247009-7247009 on Build GRCh38
rs781561894 Chr.17: 7247159-7247159 on Build GRCh38
rs893612889 Chr.17: 7247150-7247150 on Build GRCh38
rs368397693 Chr.17: 7247047-7247047 on Build GRCh38
rs150474884 Chr.17: 7247094-7247094 on Build GRCh38
rs751823609 Chr.17: 7247144-7247144 on Build GRCh38
rs1024518600 Chr.17: 7247023-7247023 on Build GRCh38
rs142495097 Chr.17: 7247172-7247172 on Build GRCh38
rs770721055 Chr.17: 7247075-7247075 on Build GRCh38
rs943690550 Chr.17: 7247004-7247004 on Build GRCh38
rs766570720 Chr.17: 7247139-7247139 on Build GRCh38
rs780325713 Chr.17: 7247032-7247032 on Build GRCh38
rs902426935 Chr.17: 7247010-7247010 on Build GRCh38
rs760550948 Chr.17: 7247096-7247096 on Build GRCh38
rs774206770 Chr.17: 7247083-7247083 on Build GRCh38
rs769302688 Chr.17: 7247056-7247056 on Build GRCh38
rs369727514 Chr.17: 7247036-7247036 on Build GRCh38
rs748655291 Chr.17: 7247051-7247051 on Build GRCh38
rs750471450 Chr.17: 7247115-7247115 on Build GRCh38

View Full Product Details