Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40843709-40843948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A6
Entrez Gene ID
1548
Gene Name
cytochrome P450 family 2 subfamily A member 6
Gene Aliases
CPA6, CYP2A, CYP2A3, CYPIIA6, P450C2A, P450PB
UniGene
Hs.250615
Gene Chromosome Location
Chr.19: 40843538-40850447 on Build GRCh38
Transcripts
NM_000762.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
213661 c.1382G>A p.R461H Chr.19: 40843899-40843899 on Build GRCh38
6151013 c.1402C>A p.P468T Chr.19: 40843879-40843879 on Build GRCh38
3990043 c.1436G>T p.G479V Chr.19: 40843845-40843845 on Build GRCh38
3534210 c.1407G>A p.K469K Chr.19: 40843874-40843874 on Build GRCh38
3726721 c.1386C>T p.L462L Chr.19: 40843895-40843895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776218213 Chr.19: 40843839-40843839 on Build GRCh38
rs752032408 Chr.19: 40843788-40843788 on Build GRCh38
rs199775494 Chr.19: 40843916-40843916 on Build GRCh38
rs28399469 Chr.19: 40843746-40843746 on Build GRCh38
rs769844210 Chr.19: 40843778-40843778 on Build GRCh38
rs1034135996 Chr.19: 40843733-40843734 on Build GRCh38
rs59322119 Chr.19: 40843748-40843748 on Build GRCh38
rs781340491 Chr.19: 40843809-40843809 on Build GRCh38
rs778526004 Chr.19: 40843799-40843799 on Build GRCh38
rs775855007 Chr.19: 40843895-40843895 on Build GRCh38
rs758261027 Chr.19: 40843794-40843794 on Build GRCh38
rs569294454 Chr.19: 40843907-40843907 on Build GRCh38
rs765005914 Chr.19: 40843842-40843842 on Build GRCh38
rs5031016 Chr.19: 40843869-40843869 on Build GRCh38
rs527558515 Chr.19: 40843756-40843756 on Build GRCh38
rs958569659 Chr.19: 40843747-40843747 on Build GRCh38
rs766075391 Chr.19: 40843904-40843904 on Build GRCh38
rs759248187 Chr.19: 40843911-40843911 on Build GRCh38
rs529455023 Chr.19: 40843868-40843868 on Build GRCh38
rs780178866 Chr.19: 40843795-40843795 on Build GRCh38
rs28399472 Chr.19: 40843737-40843737 on Build GRCh38
rs776763346 Chr.19: 40843773-40843773 on Build GRCh38
rs28399468 Chr.19: 40843827-40843827 on Build GRCh38
rs145014075 Chr.19: 40843881-40843881 on Build GRCh38
rs746173331 Chr.19: 40843888-40843888 on Build GRCh38
rs143166033 Chr.19: 40843816-40843816 on Build GRCh38
rs139835762 Chr.19: 40843799-40843799 on Build GRCh38
rs777746585 Chr.19: 40843924-40843924 on Build GRCh38
rs754911002 Chr.19: 40843802-40843802 on Build GRCh38
rs907891003 Chr.19: 40843887-40843887 on Build GRCh38
rs767844164 Chr.19: 40843836-40843836 on Build GRCh38
rs761422918 Chr.19: 40843897-40843897 on Build GRCh38
rs763617318 Chr.19: 40843789-40843789 on Build GRCh38
rs774704440 Chr.19: 40843886-40843886 on Build GRCh38
rs773644130 Chr.19: 40843781-40843781 on Build GRCh38
rs370375659 Chr.19: 40843873-40843873 on Build GRCh38
rs771101781 Chr.19: 40843885-40843885 on Build GRCh38
rs756938752 Chr.19: 40843793-40843793 on Build GRCh38
rs753849607 Chr.19: 40843926-40843926 on Build GRCh38
rs772795889 Chr.19: 40843900-40843900 on Build GRCh38
rs28399470 Chr.19: 40843742-40843742 on Build GRCh38
rs6413474 Chr.19: 40843854-40843854 on Build GRCh38
rs562378426 Chr.19: 40843731-40843731 on Build GRCh38
rs747867214 Chr.19: 40843875-40843877 on Build GRCh38
rs561053481 Chr.19: 40843858-40843858 on Build GRCh38
rs973881545 Chr.19: 40843821-40843821 on Build GRCh38
rs748123859 Chr.19: 40843814-40843814 on Build GRCh38
rs550797906 Chr.19: 40843871-40843871 on Build GRCh38
rs749536457 Chr.19: 40843884-40843884 on Build GRCh38
rs112296260 Chr.19: 40843813-40843813 on Build GRCh38
rs768823423 Chr.19: 40843765-40843765 on Build GRCh38
rs770115266 Chr.19: 40843815-40843815 on Build GRCh38
rs28399471 Chr.19: 40843739-40843739 on Build GRCh38
rs5031017 Chr.19: 40843845-40843845 on Build GRCh38
rs763293828 Chr.19: 40843782-40843782 on Build GRCh38
rs560208518 Chr.19: 40843748-40843748 on Build GRCh38
rs750010445 Chr.19: 40843844-40843844 on Build GRCh38
rs201305272 Chr.19: 40843915-40843915 on Build GRCh38
rs764629023 Chr.19: 40843899-40843899 on Build GRCh38
rs7248240 Chr.19: 40843735-40843735 on Build GRCh38
rs762723048 Chr.19: 40843901-40843901 on Build GRCh38

View Full Product Details